Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 132728442-132728915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ40288
Entrez Gene ID
286023
Gene Name
uncharacterized FLJ40288
Gene Aliases
-
UniGene
Hs.671875
Gene Chromosome Location
Chr.7: 132648794-132728769 on Build GRCh38
Transcripts
NR_046323.1

SNP Detail

SNP ID Chromosome Location
rs1014239786 Chr.7: 132728483-132728483 on Build GRCh38
rs538511174 Chr.7: 132728635-132728635 on Build GRCh38
rs1008617961 Chr.7: 132728858-132728858 on Build GRCh38
rs569366313 Chr.7: 132728603-132728603 on Build GRCh38
rs1056261989 Chr.7: 132728475-132728475 on Build GRCh38
rs984834774 Chr.7: 132728835-132728835 on Build GRCh38
rs1045691705 Chr.7: 132728503-132728503 on Build GRCh38
rs950136582 Chr.7: 132728644-132728644 on Build GRCh38
rs903483330 Chr.7: 132728508-132728508 on Build GRCh38
rs529441374 Chr.7: 132728585-132728585 on Build GRCh38
rs1055621049 Chr.7: 132728614-132728614 on Build GRCh38
rs552772023 Chr.7: 132728880-132728880 on Build GRCh38
rs894492363 Chr.7: 132728480-132728480 on Build GRCh38
rs999054330 Chr.7: 132728548-132728548 on Build GRCh38
rs999064145 Chr.7: 132728736-132728736 on Build GRCh38
rs758357484 Chr.7: 132728495-132728495 on Build GRCh38
rs952136282 Chr.7: 132728489-132728489 on Build GRCh38
rs920458420 Chr.7: 132728593-132728593 on Build GRCh38
rs1008341912 Chr.7: 132728641-132728641 on Build GRCh38
rs563854748 Chr.7: 132728827-132728827 on Build GRCh38
rs1045741560 Chr.7: 132728689-132728689 on Build GRCh38
rs546722251 Chr.7: 132728805-132728805 on Build GRCh38
rs1029130905 Chr.7: 132728572-132728572 on Build GRCh38
rs566999065 Chr.7: 132728509-132728509 on Build GRCh38
rs1010673542 Chr.7: 132728554-132728554 on Build GRCh38
rs893317742 Chr.7: 132728764-132728764 on Build GRCh38
rs964328748 Chr.7: 132728698-132728698 on Build GRCh38
rs1045665434 Chr.7: 132728588-132728588 on Build GRCh38
rs961682888 Chr.7: 132728889-132728889 on Build GRCh38
rs911552842 Chr.7: 132728584-132728584 on Build GRCh38
rs182464301 Chr.7: 132728596-132728596 on Build GRCh38
rs539487849 Chr.7: 132728809-132728809 on Build GRCh38
rs770323214 Chr.7: 132728587-132728587 on Build GRCh38
rs558492058 Chr.7: 132728655-132728655 on Build GRCh38
rs996678711 Chr.7: 132728719-132728719 on Build GRCh38
rs957729560 Chr.7: 132728553-132728553 on Build GRCh38
rs747970951 Chr.7: 132728467-132728467 on Build GRCh38
rs377505036 Chr.7: 132728851-132728851 on Build GRCh38
rs1028472524 Chr.7: 132728830-132728830 on Build GRCh38
rs747914279 Chr.7: 132728555-132728556 on Build GRCh38
rs370484632 Chr.7: 132728636-132728636 on Build GRCh38
rs903156687 Chr.7: 132728704-132728704 on Build GRCh38
rs953141992 Chr.7: 132728573-132728573 on Build GRCh38
rs952542943 Chr.7: 132728782-132728782 on Build GRCh38
rs1018624763 Chr.7: 132728860-132728860 on Build GRCh38
rs561695209 Chr.7: 132728576-132728576 on Build GRCh38
rs992608797 Chr.7: 132728867-132728867 on Build GRCh38
rs777880667 Chr.7: 132728552-132728552 on Build GRCh38
rs776265277 Chr.7: 132728589-132728589 on Build GRCh38
rs971936138 Chr.7: 132728590-132728590 on Build GRCh38
rs918199507 Chr.7: 132728564-132728564 on Build GRCh38
rs1055092204 Chr.7: 132728756-132728756 on Build GRCh38
rs948245682 Chr.7: 132728566-132728566 on Build GRCh38

View Full Product Details