Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 128008786-128009284 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR1206
Entrez Gene ID
100302170
Gene Name
microRNA 1206
Gene Aliases
MIRN1206, hsa-mir-1206
UniGene
Gene Chromosome Location
Chr.8: 128008898-128008956 on Build GRCh38
Transcripts
NR_031611.1 NR_003367.3

Gene Symbol
PVT1
Entrez Gene ID
5820
Gene Name
Pvt1 oncogene (non-protein coding)
Gene Aliases
LINC00079, MYC, NCRNA00079, onco-lncRNA-100
UniGene
Hs.133107
Gene Chromosome Location
Chr.8: 127794533-128101253 on Build GRCh38
Transcripts
NR_031611.1 NR_003367.3

SNP Detail

SNP ID Chromosome Location
rs530670370 Chr.8: 128009039-128009039 on Build GRCh38
rs916102487 Chr.8: 128008812-128008812 on Build GRCh38
rs145661347 Chr.8: 128009165-128009167 on Build GRCh38
rs879238981 Chr.8: 128009031-128009033 on Build GRCh38
rs773910631 Chr.8: 128008975-128008975 on Build GRCh38
rs751325981 Chr.8: 128008990-128008991 on Build GRCh38
rs199769002 Chr.8: 128009163-128009164 on Build GRCh38
rs762391638 Chr.8: 128008892-128008892 on Build GRCh38
rs927585563 Chr.8: 128009005-128009005 on Build GRCh38
rs369095142 Chr.8: 128008930-128008930 on Build GRCh38
rs755944050 Chr.8: 128008902-128008902 on Build GRCh38
rs761736814 Chr.8: 128009004-128009004 on Build GRCh38
rs931442387 Chr.8: 128009150-128009150 on Build GRCh38
rs369753167 Chr.8: 128008858-128008858 on Build GRCh38
rs772821618 Chr.8: 128008952-128008952 on Build GRCh38
rs377049078 Chr.8: 128008914-128008914 on Build GRCh38
rs901008562 Chr.8: 128009225-128009225 on Build GRCh38
rs192964107 Chr.8: 128008834-128008834 on Build GRCh38
rs997292482 Chr.8: 128008910-128008910 on Build GRCh38
rs377133876 Chr.8: 128008969-128008969 on Build GRCh38
rs138407144 Chr.8: 128009097-128009097 on Build GRCh38
rs763573029 Chr.8: 128008972-128008972 on Build GRCh38
rs369549987 Chr.8: 128008973-128008973 on Build GRCh38
rs2114358 Chr.8: 128008933-128008933 on Build GRCh38
rs764863860 Chr.8: 128008883-128008883 on Build GRCh38
rs1036678256 Chr.8: 128008856-128008856 on Build GRCh38
rs771337923 Chr.8: 128008965-128008965 on Build GRCh38
rs1036012506 Chr.8: 128009189-128009189 on Build GRCh38
rs762623764 Chr.8: 128008970-128008970 on Build GRCh38
rs563264501 Chr.8: 128009033-128009033 on Build GRCh38
rs949136889 Chr.8: 128009041-128009041 on Build GRCh38
rs374446759 Chr.8: 128008901-128008901 on Build GRCh38
rs200251967 Chr.8: 128008851-128008851 on Build GRCh38
rs751467648 Chr.8: 128008896-128008896 on Build GRCh38
rs774953569 Chr.8: 128008848-128008848 on Build GRCh38
rs565537095 Chr.8: 128008890-128008890 on Build GRCh38
rs748639504 Chr.8: 128008928-128008928 on Build GRCh38
rs887361336 Chr.8: 128008875-128008875 on Build GRCh38
rs777551571 Chr.8: 128008886-128008886 on Build GRCh38
rs930999952 Chr.8: 128009154-128009154 on Build GRCh38
rs908765782 Chr.8: 128009167-128009167 on Build GRCh38
rs143900669 Chr.8: 128009155-128009155 on Build GRCh38
rs370500199 Chr.8: 128008978-128008978 on Build GRCh38
rs757234576 Chr.8: 128008899-128008899 on Build GRCh38
rs755073346 Chr.8: 128008927-128008927 on Build GRCh38
rs931551976 Chr.8: 128008814-128008814 on Build GRCh38
rs759211418 Chr.8: 128008877-128008877 on Build GRCh38
rs991562701 Chr.8: 128009060-128009060 on Build GRCh38
rs1006830028 Chr.8: 128009168-128009168 on Build GRCh38
rs529134161 Chr.8: 128008811-128008811 on Build GRCh38
rs1011494439 Chr.8: 128008939-128008939 on Build GRCh38
rs889615878 Chr.8: 128008906-128008906 on Build GRCh38
rs772530326 Chr.8: 128008853-128008853 on Build GRCh38
rs187960876 Chr.8: 128008826-128008826 on Build GRCh38
rs1019414799 Chr.8: 128009195-128009195 on Build GRCh38
rs2392823 Chr.8: 128009016-128009016 on Build GRCh38
rs940286824 Chr.8: 128009176-128009176 on Build GRCh38
rs540604062 Chr.8: 128008865-128008865 on Build GRCh38
rs80050458 Chr.8: 128009148-128009148 on Build GRCh38
rs747269494 Chr.8: 128008953-128008953 on Build GRCh38
rs780084284 Chr.8: 128008912-128008912 on Build GRCh38
rs763615575 Chr.8: 128008894-128008894 on Build GRCh38

View Full Product Details