Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 19456095-19456655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCORP1
Entrez Gene ID
286554
Gene Name
BCL6 corepressor pseudogene 1
Gene Aliases
BCORL2
UniGene
Hs.211713
Gene Chromosome Location
Chr.Y: 19455431-19503153 on Build GRCh38
Transcripts
NR_002923.2 NR_033732.1

SNP Detail

SNP ID Chromosome Location
rs778667816 Chr.Y: 19456127-19456127 on Build GRCh38
rs1034813325 Chr.Y: 19456463-19456463 on Build GRCh38
rs766775933 Chr.Y: 19456598-19456598 on Build GRCh38
rs896343245 Chr.Y: 19456524-19456524 on Build GRCh38
rs1001951831 Chr.Y: 19456315-19456315 on Build GRCh38
rs754563261 Chr.Y: 19456421-19456421 on Build GRCh38

View Full Product Details