Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55746526-55747116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES1P2
Entrez Gene ID
390732
Gene Name
carboxylesterase 1 pseudogene 2
Gene Aliases
-
UniGene
Hs.554599
Gene Chromosome Location
Chr.16: 55724925-55750211 on Build GRCh38
Transcripts
NR_033740.1

SNP Detail

SNP ID Chromosome Location
rs189151512 Chr.16: 55746575-55746575 on Build GRCh38
rs116708815 Chr.16: 55746630-55746630 on Build GRCh38
rs150244253 Chr.16: 55746616-55746616 on Build GRCh38
rs560823551 Chr.16: 55746642-55746642 on Build GRCh38
rs112520198 Chr.16: 55746991-55746991 on Build GRCh38
rs572024489 Chr.16: 55746896-55746896 on Build GRCh38
rs573774112 Chr.16: 55746590-55746590 on Build GRCh38
rs138911629 Chr.16: 55746688-55746688 on Build GRCh38
rs536291190 Chr.16: 55746745-55746745 on Build GRCh38
rs753254486 Chr.16: 55747020-55747020 on Build GRCh38
rs111273487 Chr.16: 55747000-55747000 on Build GRCh38
rs547679177 Chr.16: 55746972-55746972 on Build GRCh38
rs565320734 Chr.16: 55746966-55746966 on Build GRCh38
rs538019619 Chr.16: 55746839-55746839 on Build GRCh38
rs558081718 Chr.16: 55746567-55746567 on Build GRCh38
rs560177209 Chr.16: 55746620-55746620 on Build GRCh38
rs181559687 Chr.16: 55746695-55746695 on Build GRCh38
rs548614945 Chr.16: 55746779-55746779 on Build GRCh38
rs767856381 Chr.16: 55746983-55746983 on Build GRCh38
rs565725549 Chr.16: 55746711-55746711 on Build GRCh38
rs543530804 Chr.16: 55746949-55746949 on Build GRCh38
rs550122652 Chr.16: 55746674-55746674 on Build GRCh38
rs577948963 Chr.16: 55746865-55746865 on Build GRCh38
rs148679053 Chr.16: 55747035-55747035 on Build GRCh38
rs552683039 Chr.16: 55747066-55747066 on Build GRCh38
rs531891867 Chr.16: 55746645-55746645 on Build GRCh38
rs117222216 Chr.16: 55746617-55746617 on Build GRCh38
rs532606211 Chr.16: 55746970-55746970 on Build GRCh38
rs778187662 Chr.16: 55747070-55747070 on Build GRCh38
rs537587857 Chr.16: 55746679-55746679 on Build GRCh38
rs143878305 Chr.16: 55747032-55747032 on Build GRCh38
rs556064187 Chr.16: 55746847-55746847 on Build GRCh38
rs576077986 Chr.16: 55746943-55746943 on Build GRCh38
rs141387790 Chr.16: 55747040-55747040 on Build GRCh38
rs561731769 Chr.16: 55747064-55747064 on Build GRCh38
rs142742054 Chr.16: 55746804-55746804 on Build GRCh38
rs571465038 Chr.16: 55747077-55747077 on Build GRCh38
rs572113049 Chr.16: 55746627-55746627 on Build GRCh38
rs556392344 Chr.16: 55746598-55746598 on Build GRCh38
rs186186185 Chr.16: 55746868-55746868 on Build GRCh38
rs761438064 Chr.16: 55746880-55746880 on Build GRCh38
rs141099821 Chr.16: 55746593-55746593 on Build GRCh38
rs8049324 Chr.16: 55746890-55746890 on Build GRCh38
rs572703727 Chr.16: 55746788-55746788 on Build GRCh38
rs762475580 Chr.16: 55746960-55746960 on Build GRCh38
rs6499782 Chr.16: 55746914-55746914 on Build GRCh38
rs191880423 Chr.16: 55746909-55746909 on Build GRCh38
rs760589768 Chr.16: 55746563-55746563 on Build GRCh38
rs554156609 Chr.16: 55746878-55746878 on Build GRCh38
rs746403964 Chr.16: 55746682-55746682 on Build GRCh38

View Full Product Details