Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21386154-21386708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF738
Entrez Gene ID
148203
Gene Name
zinc finger protein 738
Gene Aliases
-
UniGene
Hs.124384
Gene Chromosome Location
Chr.19: 21358933-21388582 on Build GRCh38
Transcripts
NR_027130.1

SNP Detail

SNP ID Chromosome Location
rs570894660 Chr.19: 21386512-21386512 on Build GRCh38
rs779869103 Chr.19: 21386443-21386443 on Build GRCh38
rs372717933 Chr.19: 21386222-21386222 on Build GRCh38
rs190620203 Chr.19: 21386412-21386412 on Build GRCh38
rs755109825 Chr.19: 21386538-21386538 on Build GRCh38
rs985957231 Chr.19: 21386387-21386391 on Build GRCh38
rs751762269 Chr.19: 21386505-21386505 on Build GRCh38
rs904764984 Chr.19: 21386236-21386236 on Build GRCh38
rs952038540 Chr.19: 21386351-21386351 on Build GRCh38
rs185718723 Chr.19: 21386337-21386337 on Build GRCh38
rs922641146 Chr.19: 21386524-21386524 on Build GRCh38
rs1042108262 Chr.19: 21386545-21386545 on Build GRCh38
rs190981849 Chr.19: 21386201-21386201 on Build GRCh38
rs958472176 Chr.19: 21386281-21386281 on Build GRCh38
rs867724933 Chr.19: 21386219-21386219 on Build GRCh38
rs182595920 Chr.19: 21386311-21386311 on Build GRCh38
rs904796752 Chr.19: 21386632-21386632 on Build GRCh38
rs143171350 Chr.19: 21386637-21386637 on Build GRCh38
rs758185101 Chr.19: 21386308-21386308 on Build GRCh38
rs765027866 Chr.19: 21386254-21386254 on Build GRCh38
rs538429352 Chr.19: 21386537-21386537 on Build GRCh38
rs761392260 Chr.19: 21386245-21386245 on Build GRCh38
rs910399426 Chr.19: 21386417-21386417 on Build GRCh38
rs139265884 Chr.19: 21386285-21386285 on Build GRCh38
rs1034498054 Chr.19: 21386276-21386276 on Build GRCh38
rs575063982 Chr.19: 21386669-21386669 on Build GRCh38
rs894650405 Chr.19: 21386672-21386673 on Build GRCh38
rs114299790 Chr.19: 21386229-21386229 on Build GRCh38
rs1041111829 Chr.19: 21386475-21386475 on Build GRCh38
rs552259779 Chr.19: 21386431-21386431 on Build GRCh38
rs748107530 Chr.19: 21386648-21386648 on Build GRCh38
rs11085447 Chr.19: 21386297-21386297 on Build GRCh38
rs529977104 Chr.19: 21386302-21386302 on Build GRCh38
rs1024023097 Chr.19: 21386294-21386294 on Build GRCh38
rs540353493 Chr.19: 21386180-21386180 on Build GRCh38
rs1041078737 Chr.19: 21386181-21386181 on Build GRCh38
rs972132181 Chr.19: 21386312-21386312 on Build GRCh38

View Full Product Details