Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96499216-96499762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEURL3
Entrez Gene ID
93082
Gene Name
neuralized E3 ubiquitin protein ligase 3
Gene Aliases
LINCR
UniGene
Hs.149219
Gene Chromosome Location
Chr.2: 96497643-96508157 on Build GRCh38
Transcripts
NM_001285485.1 NM_001285486.1 NR_104346.1 XM_011512178.2 XM_011512179.2

SNP Detail

SNP ID Chromosome Location
rs956537983 Chr.2: 96499346-96499346 on Build GRCh38
rs1026267471 Chr.2: 96499733-96499733 on Build GRCh38
rs930122949 Chr.2: 96499501-96499501 on Build GRCh38
rs778012940 Chr.2: 96499423-96499423 on Build GRCh38
rs997431175 Chr.2: 96499728-96499728 on Build GRCh38
rs747248029 Chr.2: 96499497-96499497 on Build GRCh38
rs919964137 Chr.2: 96499675-96499675 on Build GRCh38
rs926541960 Chr.2: 96499608-96499608 on Build GRCh38
rs78987811 Chr.2: 96499482-96499482 on Build GRCh38
rs999395934 Chr.2: 96499395-96499395 on Build GRCh38
rs1047171472 Chr.2: 96499730-96499730 on Build GRCh38
rs375141804 Chr.2: 96499435-96499435 on Build GRCh38
rs980719188 Chr.2: 96499436-96499436 on Build GRCh38
rs562271656 Chr.2: 96499307-96499307 on Build GRCh38
rs755041441 Chr.2: 96499495-96499496 on Build GRCh38
rs924318591 Chr.2: 96499540-96499540 on Build GRCh38
rs765826178 Chr.2: 96499490-96499490 on Build GRCh38
rs756583493 Chr.2: 96499378-96499378 on Build GRCh38
rs777959169 Chr.2: 96499493-96499493 on Build GRCh38
rs868651567 Chr.2: 96499344-96499344 on Build GRCh38
rs899462429 Chr.2: 96499534-96499534 on Build GRCh38
rs145836227 Chr.2: 96499713-96499713 on Build GRCh38
rs751336677 Chr.2: 96499584-96499584 on Build GRCh38
rs116087310 Chr.2: 96499345-96499345 on Build GRCh38
rs373790377 Chr.2: 96499508-96499508 on Build GRCh38
rs746803128 Chr.2: 96499425-96499425 on Build GRCh38
rs966000726 Chr.2: 96499457-96499458 on Build GRCh38
rs373508867 Chr.2: 96499377-96499377 on Build GRCh38
rs1039239784 Chr.2: 96499683-96499683 on Build GRCh38
rs1032067854 Chr.2: 96499541-96499541 on Build GRCh38
rs527548598 Chr.2: 96499369-96499369 on Build GRCh38
rs577683462 Chr.2: 96499635-96499635 on Build GRCh38
rs769306737 Chr.2: 96499439-96499439 on Build GRCh38
rs902118455 Chr.2: 96499716-96499716 on Build GRCh38
rs980674232 Chr.2: 96499607-96499607 on Build GRCh38
rs752746126 Chr.2: 96499491-96499491 on Build GRCh38
rs775160103 Chr.2: 96499443-96499443 on Build GRCh38
rs534971667 Chr.2: 96499509-96499509 on Build GRCh38
rs1056149028 Chr.2: 96499281-96499281 on Build GRCh38
rs766688250 Chr.2: 96499736-96499736 on Build GRCh38
rs577794697 Chr.2: 96499644-96499644 on Build GRCh38
rs1017929952 Chr.2: 96499284-96499284 on Build GRCh38
rs370874357 Chr.2: 96499365-96499365 on Build GRCh38
rs759074426 Chr.2: 96499340-96499340 on Build GRCh38
rs773215151 Chr.2: 96499469-96499469 on Build GRCh38
rs888209870 Chr.2: 96499724-96499724 on Build GRCh38
rs1040359957 Chr.2: 96499521-96499521 on Build GRCh38
rs934425880 Chr.2: 96499553-96499553 on Build GRCh38
rs767514326 Chr.2: 96499496-96499496 on Build GRCh38
rs555832099 Chr.2: 96499740-96499740 on Build GRCh38
rs758541255 Chr.2: 96499492-96499492 on Build GRCh38
rs941284313 Chr.2: 96499260-96499260 on Build GRCh38
rs772264428 Chr.2: 96499421-96499421 on Build GRCh38
rs189497677 Chr.2: 96499656-96499656 on Build GRCh38
rs762600543 Chr.2: 96499463-96499463 on Build GRCh38
rs979163483 Chr.2: 96499329-96499329 on Build GRCh38
rs964087873 Chr.2: 96499288-96499288 on Build GRCh38
rs558064547 Chr.2: 96499516-96499516 on Build GRCh38
rs780602929 Chr.2: 96499385-96499385 on Build GRCh38
rs115561660 Chr.2: 96499241-96499241 on Build GRCh38
rs533188042 Chr.2: 96499420-96499420 on Build GRCh38
rs1024807048 Chr.2: 96499591-96499591 on Build GRCh38
rs930050967 Chr.2: 96499729-96499729 on Build GRCh38
rs763805880 Chr.2: 96499470-96499470 on Build GRCh38
rs547694357 Chr.2: 96499370-96499370 on Build GRCh38
rs927313578 Chr.2: 96499466-96499466 on Build GRCh38
rs970590440 Chr.2: 96499592-96499592 on Build GRCh38
rs370779737 Chr.2: 96499434-96499434 on Build GRCh38
rs541503396 Chr.2: 96499356-96499356 on Build GRCh38
rs974500825 Chr.2: 96499638-96499638 on Build GRCh38
rs774210783 Chr.2: 96499473-96499473 on Build GRCh38
rs149375652 Chr.2: 96499481-96499481 on Build GRCh38
rs1000952912 Chr.2: 96499535-96499535 on Build GRCh38
rs778894245 Chr.2: 96499401-96499401 on Build GRCh38
rs956424352 Chr.2: 96499568-96499568 on Build GRCh38
rs893334989 Chr.2: 96499408-96499408 on Build GRCh38
rs376338833 Chr.2: 96499391-96499391 on Build GRCh38
rs879252895 Chr.2: 96499502-96499502 on Build GRCh38
rs1009347461 Chr.2: 96499575-96499575 on Build GRCh38
rs570630004 Chr.2: 96499393-96499393 on Build GRCh38

View Full Product Details