Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 42511472-42512030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRP7A
Entrez Gene ID
27341
Gene Name
ribosomal RNA processing 7 homolog A
Gene Aliases
BK126B4.3, CGI-96
UniGene
Hs.745006
Gene Chromosome Location
Chr.22: 42508335-42519823 on Build GRCh38
Transcripts
NM_015703.4 NR_027786.1

Gene Symbol
SERHL
Entrez Gene ID
94009
Gene Name
serine hydrolase-like (pseudogene)
Gene Aliases
BK126B4.1, BK126B4.2, HS126B42, dJ222E13.1
UniGene
Hs.360940
Gene Chromosome Location
Chr.22: 42500579-42512560 on Build GRCh38
Transcripts
NM_015703.4 NR_027786.1

SNP Detail

SNP ID Chromosome Location
rs970544426 Chr.22: 42511552-42511552 on Build GRCh38
rs117727340 Chr.22: 42511543-42511543 on Build GRCh38
rs572981166 Chr.22: 42511654-42511654 on Build GRCh38
rs1023504134 Chr.22: 42511729-42511729 on Build GRCh38
rs987571758 Chr.22: 42511864-42511864 on Build GRCh38
rs535814418 Chr.22: 42511680-42511680 on Build GRCh38
rs10428047 Chr.22: 42511857-42511857 on Build GRCh38
rs558788804 Chr.22: 42511959-42511959 on Build GRCh38
rs1051810901 Chr.22: 42511511-42511511 on Build GRCh38
rs142093321 Chr.22: 42511741-42511741 on Build GRCh38
rs924112126 Chr.22: 42512004-42512004 on Build GRCh38
rs775475843 Chr.22: 42511696-42511707 on Build GRCh38
rs933471044 Chr.22: 42511694-42511694 on Build GRCh38
rs1005249822 Chr.22: 42511797-42511797 on Build GRCh38
rs544450174 Chr.22: 42511759-42511759 on Build GRCh38
rs372483931 Chr.22: 42511762-42511762 on Build GRCh38
rs1005302087 Chr.22: 42511939-42511939 on Build GRCh38
rs906424242 Chr.22: 42511919-42511919 on Build GRCh38
rs533989657 Chr.22: 42511927-42511927 on Build GRCh38
rs1046917028 Chr.22: 42511932-42511932 on Build GRCh38
rs191545077 Chr.22: 42511790-42511790 on Build GRCh38
rs1016643838 Chr.22: 42511940-42511940 on Build GRCh38
rs140140261 Chr.22: 42511586-42511586 on Build GRCh38
rs1032239142 Chr.22: 42511540-42511540 on Build GRCh38
rs747905018 Chr.22: 42511520-42511520 on Build GRCh38
rs954971171 Chr.22: 42511982-42511982 on Build GRCh38
rs150061568 Chr.22: 42511853-42511853 on Build GRCh38
rs552307324 Chr.22: 42511824-42511824 on Build GRCh38
rs915221328 Chr.22: 42511903-42511903 on Build GRCh38
rs540349835 Chr.22: 42511717-42511717 on Build GRCh38
rs1029671967 Chr.22: 42511963-42511963 on Build GRCh38
rs893749472 Chr.22: 42511723-42511723 on Build GRCh38
rs143497752 Chr.22: 42511608-42511608 on Build GRCh38
rs1045388492 Chr.22: 42511913-42511913 on Build GRCh38
rs933382054 Chr.22: 42511503-42511503 on Build GRCh38
rs12628879 Chr.22: 42511898-42511898 on Build GRCh38
rs902530623 Chr.22: 42511527-42511527 on Build GRCh38
rs534853944 Chr.22: 42511536-42511536 on Build GRCh38
rs577317495 Chr.22: 42512006-42512006 on Build GRCh38
rs935348168 Chr.22: 42511707-42511707 on Build GRCh38
rs184263709 Chr.22: 42511611-42511611 on Build GRCh38
rs951027678 Chr.22: 42511602-42511602 on Build GRCh38
rs558877929 Chr.22: 42511730-42511730 on Build GRCh38
rs374789246 Chr.22: 42511665-42511665 on Build GRCh38
rs1053704199 Chr.22: 42511713-42511713 on Build GRCh38
rs139433212 Chr.22: 42511825-42511825 on Build GRCh38
rs999444631 Chr.22: 42511529-42511529 on Build GRCh38
rs922090643 Chr.22: 42511687-42511687 on Build GRCh38
rs569580546 Chr.22: 42511596-42511596 on Build GRCh38
rs886880840 Chr.22: 42511934-42511934 on Build GRCh38
rs1012576109 Chr.22: 42511728-42511728 on Build GRCh38
rs61752533 Chr.22: 42511893-42511893 on Build GRCh38
rs1023873575 Chr.22: 42511549-42511549 on Build GRCh38
rs964053076 Chr.22: 42511943-42511943 on Build GRCh38
rs566739397 Chr.22: 42511899-42511899 on Build GRCh38
rs530046766 Chr.22: 42511763-42511763 on Build GRCh38
rs187091616 Chr.22: 42511738-42511738 on Build GRCh38
rs948043802 Chr.22: 42511910-42511910 on Build GRCh38
rs906365561 Chr.22: 42511748-42511748 on Build GRCh38
rs12485160 Chr.22: 42511613-42511613 on Build GRCh38
rs996503170 Chr.22: 42511960-42511960 on Build GRCh38
rs150907835 Chr.22: 42511775-42511775 on Build GRCh38
rs930293571 Chr.22: 42511720-42511720 on Build GRCh38
rs557093945 Chr.22: 42511812-42511812 on Build GRCh38
rs924056795 Chr.22: 42511869-42511869 on Build GRCh38
rs983762683 Chr.22: 42511612-42511612 on Build GRCh38
rs560408947 Chr.22: 42511788-42511788 on Build GRCh38
rs987625017 Chr.22: 42511989-42511989 on Build GRCh38
rs1051800965 Chr.22: 42511696-42511699 on Build GRCh38
rs558079000 Chr.22: 42511915-42511915 on Build GRCh38
rs767776553 Chr.22: 42511534-42511534 on Build GRCh38
rs539276778 Chr.22: 42511544-42511544 on Build GRCh38
rs5758775 Chr.22: 42511851-42511851 on Build GRCh38

View Full Product Details