Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101353111-101353285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTK
Entrez Gene ID
695
Gene Name
Bruton tyrosine kinase
Gene Aliases
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
UniGene
Hs.159494
Gene Chromosome Location
Chr.X: 101349447-101390796 on Build GRCh38
Transcripts
NM_000061.2 NM_001287344.1 NM_001287345.1

SNP Detail

SNP ID Chromosome Location
rs200762027 Chr.X: 101353167-101353167 on Build GRCh38
rs3027646 Chr.X: 101353220-101353220 on Build GRCh38
rs372424571 Chr.X: 101353170-101353170 on Build GRCh38
rs128621209 Chr.X: 101353264-101353264 on Build GRCh38
rs1010302002 Chr.X: 101353176-101353176 on Build GRCh38
rs782244933 Chr.X: 101353249-101353249 on Build GRCh38
rs371003927 Chr.X: 101353138-101353138 on Build GRCh38
rs128622211 Chr.X: 101353196-101353196 on Build GRCh38
rs1057287168 Chr.X: 101353162-101353162 on Build GRCh38
rs781794942 Chr.X: 101353146-101353146 on Build GRCh38
rs782609610 Chr.X: 101353189-101353189 on Build GRCh38
rs128621210 Chr.X: 101353213-101353213 on Build GRCh38
rs782503550 Chr.X: 101353218-101353218 on Build GRCh38
rs782283931 Chr.X: 101353242-101353242 on Build GRCh38
rs1135363 Chr.X: 101353203-101353203 on Build GRCh38

View Full Product Details