Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 64950682-64950856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH11
Entrez Gene ID
1009
Gene Name
cadherin 11
Gene Aliases
CAD11, CDHOB, OB, OSF-4
UniGene
Hs.116471
Gene Chromosome Location
Chr.16: 64943753-65126112 on Build GRCh38
Transcripts
NM_001308392.1 NM_001797.3 XM_005255761.3 XM_005255762.2 XM_005255763.2 XM_011522803.2 XM_017022830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4871464 c.1894G>T p.G632C Chr.16: 64950767-64950767 on Build GRCh38
2993391 c.1814A>T p.N605I Chr.16: 64950847-64950847 on Build GRCh38
2993389 c.1873G>A p.A625T Chr.16: 64950788-64950788 on Build GRCh38
7306712 c.1869C>A p.I623I Chr.16: 64950792-64950792 on Build GRCh38
6190914 c.1845G>A p.L615L Chr.16: 64950816-64950816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745525216 Chr.16: 64950783-64950783 on Build GRCh38
rs756786724 Chr.16: 64950775-64950775 on Build GRCh38
rs556733053 Chr.16: 64950758-64950758 on Build GRCh38
rs766525925 Chr.16: 64950830-64950830 on Build GRCh38
rs767585778 Chr.16: 64950725-64950725 on Build GRCh38
rs767603767 Chr.16: 64950812-64950812 on Build GRCh38
rs764709799 Chr.16: 64950720-64950720 on Build GRCh38
rs932110627 Chr.16: 64950708-64950708 on Build GRCh38
rs774503953 Chr.16: 64950721-64950721 on Build GRCh38
rs905543757 Chr.16: 64950835-64950835 on Build GRCh38
rs572049132 Chr.16: 64950736-64950736 on Build GRCh38
rs1051867226 Chr.16: 64950718-64950718 on Build GRCh38
rs368601615 Chr.16: 64950744-64950744 on Build GRCh38
rs372527203 Chr.16: 64950807-64950807 on Build GRCh38
rs981447409 Chr.16: 64950710-64950710 on Build GRCh38
rs766393142 Chr.16: 64950744-64950745 on Build GRCh38
rs560953034 Chr.16: 64950809-64950809 on Build GRCh38
rs780620859 Chr.16: 64950779-64950779 on Build GRCh38
rs529917059 Chr.16: 64950837-64950837 on Build GRCh38
rs760939414 Chr.16: 64950818-64950818 on Build GRCh38
rs752547668 Chr.16: 64950747-64950747 on Build GRCh38
rs775862772 Chr.16: 64950794-64950794 on Build GRCh38
rs752425143 Chr.16: 64950831-64950831 on Build GRCh38
rs773232522 Chr.16: 64950817-64950817 on Build GRCh38
rs763021806 Chr.16: 64950732-64950732 on Build GRCh38
rs201311098 Chr.16: 64950728-64950728 on Build GRCh38
rs866544694 Chr.16: 64950798-64950798 on Build GRCh38
rs762014357 Chr.16: 64950723-64950723 on Build GRCh38
rs749671969 Chr.16: 64950800-64950800 on Build GRCh38
rs541059450 Chr.16: 64950737-64950737 on Build GRCh38
rs758223898 Chr.16: 64950833-64950833 on Build GRCh38
rs777694874 Chr.16: 64950760-64950760 on Build GRCh38
rs769244467 Chr.16: 64950806-64950806 on Build GRCh38
rs765077367 Chr.16: 64950739-64950739 on Build GRCh38
rs758275194 Chr.16: 64950748-64950748 on Build GRCh38
rs769387085 Chr.16: 64950788-64950788 on Build GRCh38

View Full Product Details