Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101349839-101350002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTK
Entrez Gene ID
695
Gene Name
Bruton tyrosine kinase
Gene Aliases
AGMX1, AT, ATK, BPK, IMD1, PSCTK1, XLA
UniGene
Hs.159494
Gene Chromosome Location
Chr.X: 101349447-101390796 on Build GRCh38
Transcripts
NM_000061.2 NM_001287344.1 NM_001287345.1

SNP Detail

SNP ID Chromosome Location
rs781952338 Chr.X: 101349948-101349948 on Build GRCh38
rs782396573 Chr.X: 101349867-101349867 on Build GRCh38
rs782709592 Chr.X: 101349900-101349900 on Build GRCh38
rs782702231 Chr.X: 101349965-101349965 on Build GRCh38
rs41300892 Chr.X: 101349940-101349940 on Build GRCh38
rs41311721 Chr.X: 101349864-101349864 on Build GRCh38
rs377028655 Chr.X: 101349874-101349874 on Build GRCh38
rs782119510 Chr.X: 101349865-101349865 on Build GRCh38
rs782047787 Chr.X: 101349888-101349888 on Build GRCh38
rs782777227 Chr.X: 101349974-101349974 on Build GRCh38
rs128622212 Chr.X: 101349910-101349910 on Build GRCh38

View Full Product Details