Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 64947988-64948141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH11
Entrez Gene ID
1009
Gene Name
cadherin 11
Gene Aliases
CAD11, CDHOB, OB, OSF-4
UniGene
Hs.116471
Gene Chromosome Location
Chr.16: 64943753-65126112 on Build GRCh38
Transcripts
NM_001308392.1 NM_001797.3 XM_005255761.3 XM_005255762.2 XM_005255763.2 XM_011522803.2 XM_017022830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4647948 c.1918C>A p.L640M Chr.16: 64948076-64948076 on Build GRCh38
4972979 c.1966G>A p.V656I Chr.16: 64948028-64948028 on Build GRCh38
239248 c.1969C>T p.R657C Chr.16: 64948025-64948025 on Build GRCh38
1200413 c.1940C>A p.P647Q Chr.16: 64948054-64948054 on Build GRCh38
6053578 c.1990G>T p.D664Y Chr.16: 64948004-64948004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763749844 Chr.16: 64948024-64948024 on Build GRCh38
rs779101151 Chr.16: 64948093-64948093 on Build GRCh38
rs748289360 Chr.16: 64948105-64948105 on Build GRCh38
rs749326608 Chr.16: 64948052-64948052 on Build GRCh38
rs992269257 Chr.16: 64948102-64948102 on Build GRCh38
rs1022367726 Chr.16: 64948043-64948043 on Build GRCh38
rs750843398 Chr.16: 64948047-64948047 on Build GRCh38
rs762560368 Chr.16: 64948018-64948018 on Build GRCh38
rs767767791 Chr.16: 64948044-64948044 on Build GRCh38
rs146502017 Chr.16: 64948050-64948050 on Build GRCh38
rs756971514 Chr.16: 64948037-64948037 on Build GRCh38
rs768421766 Chr.16: 64948030-64948032 on Build GRCh38
rs755036111 Chr.16: 64948054-64948054 on Build GRCh38
rs573271692 Chr.16: 64948110-64948110 on Build GRCh38
rs960872773 Chr.16: 64948067-64948067 on Build GRCh38
rs201059793 Chr.16: 64948049-64948049 on Build GRCh38
rs751145538 Chr.16: 64948028-64948028 on Build GRCh38

View Full Product Details