Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37223801-37224038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPN2
Entrez Gene ID
6185
Gene Name
ribophorin II
Gene Aliases
RIBIIR, RPN-II, RPNII, SWP1
UniGene
Hs.370895
Gene Chromosome Location
Chr.20: 37179046-37241623 on Build GRCh38
Transcripts
NM_001135771.2 NM_001324299.1 NM_001324301.1 NM_001324302.1 NM_001324303.1 NM_001324304.1 NM_001324305.1 NM_001324306.1 NM_002951.4 XM_006723851.2 XM_006723852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5930035 c.1172C>T p.P391L Chr.20: 37223957-37223957 on Build GRCh38
4098134 c.1096A>G p.R366G Chr.20: 37223881-37223881 on Build GRCh38
5814730 c.1096A>T p.R366* Chr.20: 37223881-37223881 on Build GRCh38
7429753 c.1109C>G p.S370C Chr.20: 37223894-37223894 on Build GRCh38
5857808 c.1184+1G>A p.? Chr.20: 37223970-37223970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367758574 Chr.20: 37223932-37223932 on Build GRCh38
rs908136417 Chr.20: 37223908-37223908 on Build GRCh38
rs769347765 Chr.20: 37223887-37223887 on Build GRCh38
rs990439291 Chr.20: 37223840-37223840 on Build GRCh38
rs752594863 Chr.20: 37223836-37223836 on Build GRCh38
rs959039239 Chr.20: 37223834-37223834 on Build GRCh38
rs933413201 Chr.20: 37223991-37223991 on Build GRCh38
rs750857139 Chr.20: 37223856-37223856 on Build GRCh38
rs760770153 Chr.20: 37223841-37223841 on Build GRCh38
rs371041505 Chr.20: 37223866-37223866 on Build GRCh38
rs149499238 Chr.20: 37223921-37223921 on Build GRCh38
rs759361933 Chr.20: 37223914-37223914 on Build GRCh38
rs373623696 Chr.20: 37223923-37223923 on Build GRCh38
rs758847133 Chr.20: 37223993-37223993 on Build GRCh38
rs2076397 Chr.20: 37223992-37223992 on Build GRCh38
rs372015507 Chr.20: 37223969-37223969 on Build GRCh38
rs748815752 Chr.20: 37223902-37223902 on Build GRCh38
rs753924496 Chr.20: 37223947-37223947 on Build GRCh38
rs886610800 Chr.20: 37224015-37224015 on Build GRCh38
rs943388232 Chr.20: 37223954-37223954 on Build GRCh38
rs780495871 Chr.20: 37223997-37223997 on Build GRCh38
rs747460123 Chr.20: 37223876-37223876 on Build GRCh38
rs937317711 Chr.20: 37223948-37223948 on Build GRCh38
rs548663185 Chr.20: 37223846-37223846 on Build GRCh38
rs138101348 Chr.20: 37223890-37223890 on Build GRCh38
rs757490362 Chr.20: 37223845-37223845 on Build GRCh38
rs117475222 Chr.20: 37223906-37223906 on Build GRCh38
rs777797703 Chr.20: 37224012-37224012 on Build GRCh38
rs6104133 Chr.20: 37224006-37224006 on Build GRCh38
rs190926021 Chr.20: 37223985-37223985 on Build GRCh38
rs553191939 Chr.20: 37223968-37223968 on Build GRCh38
rs770535836 Chr.20: 37223903-37223903 on Build GRCh38
rs780728799 Chr.20: 37223874-37223874 on Build GRCh38
rs754094718 Chr.20: 37223843-37223843 on Build GRCh38
rs372128801 Chr.20: 37224001-37224001 on Build GRCh38
rs775420634 Chr.20: 37223931-37223931 on Build GRCh38
rs977981583 Chr.20: 37223885-37223885 on Build GRCh38
rs201745779 Chr.20: 37223842-37223842 on Build GRCh38

View Full Product Details