Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37179286-37179527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH8
Entrez Gene ID
140699
Gene Name
maestro heat like repeat family member 8
Gene Aliases
C20orf131, C20orf132, dJ621N11.3, dJ621N11.4
UniGene
Hs.349125
Gene Chromosome Location
Chr.20: 37101226-37179588 on Build GRCh38
Transcripts
NM_152503.5 NM_213631.2 NM_213632.2 NM_001135771.2 NM_001324299.1 NM_001324301.1 NM_001324302.1 NM_001324303.1 NM_001324304.1 NM_001324305.1 NM_001324306.1 NM_002951.4 XM_006723851.2 XM_006723852.2

Gene Symbol
RPN2
Entrez Gene ID
6185
Gene Name
ribophorin II
Gene Aliases
RIBIIR, RPN-II, RPNII, SWP1
UniGene
Hs.370895
Gene Chromosome Location
Chr.20: 37179046-37241623 on Build GRCh38
Transcripts
NM_152503.5 NM_213631.2 NM_213632.2 NM_001135771.2 NM_001324299.1 NM_001324301.1 NM_001324302.1 NM_001324303.1 NM_001324304.1 NM_001324305.1 NM_001324306.1 NM_002951.4 XM_006723851.2 XM_006723852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1592567 c.93_93+1ins29 p.H32fs*60 Chr.20: 37179387-37179388 on Build GRCh38
6498875 c.147G>A p.E49E Chr.20: 37179333-37179333 on Build GRCh38
4134505 c.174C>G p.S58R Chr.20: 37179386-37179386 on Build GRCh38
5445854 c.90_91ins29 p.H32fs*60 Chr.20: 37179390-37179391 on Build GRCh38
5445853 c.169_170ins29 p.S58fs*61 Chr.20: 37179390-37179391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763402609 Chr.20: 37179385-37179386 on Build GRCh38
rs752600317 Chr.20: 37179382-37179382 on Build GRCh38
rs976592202 Chr.20: 37179479-37179479 on Build GRCh38
rs971972758 Chr.20: 37179368-37179368 on Build GRCh38
rs777314104 Chr.20: 37179461-37179461 on Build GRCh38
rs753743659 Chr.20: 37179391-37179391 on Build GRCh38
rs11467214 Chr.20: 37179387-37179388 on Build GRCh38
rs755846009 Chr.20: 37179384-37179384 on Build GRCh38
rs199988728 Chr.20: 37179431-37179431 on Build GRCh38
rs904737400 Chr.20: 37179480-37179480 on Build GRCh38
rs751282909 Chr.20: 37179330-37179330 on Build GRCh38
rs529171256 Chr.20: 37179449-37179449 on Build GRCh38
rs757358667 Chr.20: 37179395-37179395 on Build GRCh38
rs961958210 Chr.20: 37179345-37179345 on Build GRCh38
rs746082220 Chr.20: 37179397-37179397 on Build GRCh38
rs780127488 Chr.20: 37179413-37179413 on Build GRCh38
rs767218190 Chr.20: 37179367-37179367 on Build GRCh38
rs762364186 Chr.20: 37179463-37179463 on Build GRCh38
rs768133019 Chr.20: 37179388-37179389 on Build GRCh38
rs917442312 Chr.20: 37179458-37179458 on Build GRCh38
rs758211713 Chr.20: 37179391-37179392 on Build GRCh38
rs1008767025 Chr.20: 37179307-37179307 on Build GRCh38
rs1003688063 Chr.20: 37179343-37179343 on Build GRCh38
rs756124820 Chr.20: 37179391-37179395 on Build GRCh38
rs759168006 Chr.20: 37179366-37179366 on Build GRCh38
rs1044799186 Chr.20: 37179319-37179319 on Build GRCh38
rs984486891 Chr.20: 37179408-37179408 on Build GRCh38
rs57415986 Chr.20: 37179390-37179391 on Build GRCh38
rs546768849 Chr.20: 37179377-37179377 on Build GRCh38
rs150766053 Chr.20: 37179362-37179363 on Build GRCh38
rs1050507778 Chr.20: 37179499-37179499 on Build GRCh38
rs909801781 Chr.20: 37179501-37179501 on Build GRCh38
rs868249284 Chr.20: 37179386-37179387 on Build GRCh38
rs935720077 Chr.20: 37179495-37179495 on Build GRCh38
rs545071843 Chr.20: 37179395-37179396 on Build GRCh38
rs952720487 Chr.20: 37179386-37179386 on Build GRCh38
rs1018719630 Chr.20: 37179342-37179342 on Build GRCh38
rs528617599 Chr.20: 37179337-37179337 on Build GRCh38
rs904962978 Chr.20: 37179322-37179322 on Build GRCh38
rs868411764 Chr.20: 37179393-37179393 on Build GRCh38
rs1130679 Chr.20: 37179315-37179315 on Build GRCh38
rs1035628911 Chr.20: 37179350-37179350 on Build GRCh38
rs1042002391 Chr.20: 37179471-37179471 on Build GRCh38
rs867759436 Chr.20: 37179392-37179392 on Build GRCh38
rs922466866 Chr.20: 37179492-37179492 on Build GRCh38
rs765949800 Chr.20: 37179326-37179326 on Build GRCh38
rs550614971 Chr.20: 37179491-37179491 on Build GRCh38
rs888901039 Chr.20: 37179506-37179506 on Build GRCh38
rs927437887 Chr.20: 37179418-37179418 on Build GRCh38

View Full Product Details