Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4698330-4698599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS3
Entrez Gene ID
124401
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Gene Aliases
-
UniGene
Hs.654859
Gene Chromosome Location
Chr.16: 4696510-4734377 on Build GRCh38
Transcripts
NM_001242929.1 NM_001308089.1 NM_001324129.1 NM_001324130.1 NM_133450.3 NR_040252.1 XM_005255119.3 XM_005255121.3 XM_011522372.1 XM_011522373.1 XM_011522374.1 XM_011522378.1 XM_017022935.1 XM_017022936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860813 c.1723C>T p.R575* Chr.16: 4698428-4698428 on Build GRCh38
6370417 c.1591C>T p.Q531* Chr.16: 4698560-4698560 on Build GRCh38
4470516 c.1665C>T p.A555A Chr.16: 4698486-4698486 on Build GRCh38
5589660 c.1650C>T p.R550R Chr.16: 4698501-4698501 on Build GRCh38
3948554 c.1611C>T p.A537A Chr.16: 4698540-4698540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761787654 Chr.16: 4698478-4698478 on Build GRCh38
rs757612903 Chr.16: 4698510-4698510 on Build GRCh38
rs762828564 Chr.16: 4698500-4698500 on Build GRCh38
rs902828549 Chr.16: 4698495-4698495 on Build GRCh38
rs770796337 Chr.16: 4698555-4698555 on Build GRCh38
rs754255284 Chr.16: 4698509-4698509 on Build GRCh38
rs377174585 Chr.16: 4698577-4698577 on Build GRCh38
rs753090421 Chr.16: 4698451-4698451 on Build GRCh38
rs184430763 Chr.16: 4698375-4698375 on Build GRCh38
rs929947683 Chr.16: 4698545-4698545 on Build GRCh38
rs781202313 Chr.16: 4698479-4698479 on Build GRCh38
rs566391799 Chr.16: 4698503-4698503 on Build GRCh38
rs369097147 Chr.16: 4698543-4698543 on Build GRCh38
rs774658340 Chr.16: 4698560-4698560 on Build GRCh38
rs1041057588 Chr.16: 4698420-4698420 on Build GRCh38
rs751255198 Chr.16: 4698579-4698579 on Build GRCh38
rs774373533 Chr.16: 4698502-4698502 on Build GRCh38
rs79936810 Chr.16: 4698456-4698456 on Build GRCh38
rs911257251 Chr.16: 4698410-4698410 on Build GRCh38
rs888600183 Chr.16: 4698488-4698488 on Build GRCh38
rs202242625 Chr.16: 4698574-4698574 on Build GRCh38
rs998284204 Chr.16: 4698534-4698534 on Build GRCh38
rs765065538 Chr.16: 4698396-4698396 on Build GRCh38
rs936519987 Chr.16: 4698415-4698415 on Build GRCh38
rs779663025 Chr.16: 4698471-4698471 on Build GRCh38
rs759639498 Chr.16: 4698563-4698563 on Build GRCh38
rs958402938 Chr.16: 4698357-4698357 on Build GRCh38
rs754537376 Chr.16: 4698531-4698531 on Build GRCh38
rs771350428 Chr.16: 4698501-4698501 on Build GRCh38
rs766686749 Chr.16: 4698428-4698428 on Build GRCh38
rs986085032 Chr.16: 4698388-4698388 on Build GRCh38
rs199765749 Chr.16: 4698484-4698484 on Build GRCh38
rs199731997 Chr.16: 4698542-4698542 on Build GRCh38
rs772304435 Chr.16: 4698564-4698564 on Build GRCh38
rs369685757 Chr.16: 4698382-4698382 on Build GRCh38
rs970997959 Chr.16: 4698369-4698369 on Build GRCh38
rs746596153 Chr.16: 4698472-4698472 on Build GRCh38
rs187447720 Chr.16: 4698378-4698378 on Build GRCh38
rs550293447 Chr.16: 4698418-4698418 on Build GRCh38
rs965387864 Chr.16: 4698358-4698358 on Build GRCh38
rs751113006 Chr.16: 4698515-4698515 on Build GRCh38
rs201289279 Chr.16: 4698457-4698457 on Build GRCh38
rs767449077 Chr.16: 4698505-4698505 on Build GRCh38
rs574932957 Chr.16: 4698566-4698566 on Build GRCh38
rs755011963 Chr.16: 4698443-4698443 on Build GRCh38
rs1046449584 Chr.16: 4698460-4698460 on Build GRCh38
rs768697604 Chr.16: 4698381-4698381 on Build GRCh38
rs909831490 Chr.16: 4698571-4698573 on Build GRCh38
rs761207585 Chr.16: 4698506-4698506 on Build GRCh38
rs764558382 Chr.16: 4698508-4698508 on Build GRCh38
rs765468904 Chr.16: 4698578-4698578 on Build GRCh38
rs1056063741 Chr.16: 4698576-4698576 on Build GRCh38
rs761672053 Chr.16: 4698393-4698393 on Build GRCh38
rs754638570 Chr.16: 4698580-4698580 on Build GRCh38
rs1023970784 Chr.16: 4698365-4698365 on Build GRCh38
rs140897507 Chr.16: 4698540-4698540 on Build GRCh38
rs758091213 Chr.16: 4698467-4698467 on Build GRCh38
rs751719693 Chr.16: 4698429-4698429 on Build GRCh38
rs767983204 Chr.16: 4698474-4698474 on Build GRCh38
rs942796817 Chr.16: 4698414-4698414 on Build GRCh38
rs763307507 Chr.16: 4698427-4698427 on Build GRCh38
rs780608870 Chr.16: 4698539-4698539 on Build GRCh38
rs749618193 Chr.16: 4698458-4698458 on Build GRCh38
rs373499350 Chr.16: 4698404-4698404 on Build GRCh38
rs879119223 Chr.16: 4698476-4698476 on Build GRCh38
rs769824676 Chr.16: 4698485-4698485 on Build GRCh38
rs977197476 Chr.16: 4698373-4698373 on Build GRCh38
rs773191081 Chr.16: 4698487-4698487 on Build GRCh38
rs754276102 Chr.16: 4698575-4698575 on Build GRCh38
rs768076620 Chr.16: 4698444-4698444 on Build GRCh38
rs760727197 Chr.16: 4698568-4698568 on Build GRCh38

View Full Product Details