Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73902297-73902562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTG2
Entrez Gene ID
72
Gene Name
actin, gamma 2, smooth muscle, enteric
Gene Aliases
ACT, ACTA3, ACTE, ACTL3, ACTSG, VSCM
UniGene
Hs.516105
Gene Chromosome Location
Chr.2: 73892966-73919653 on Build GRCh38
Transcripts
NM_001199893.1 NM_001615.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6158866 c.202C>G p.L68V Chr.2: 73902435-73902435 on Build GRCh38
6158867 c.135G>T p.M45I Chr.2: 73902368-73902368 on Build GRCh38
4532268 c.186G>A p.K62K Chr.2: 73902419-73902419 on Build GRCh38
5397395 c.225C>T p.G75G Chr.2: 73902458-73902458 on Build GRCh38
4532496 c.188G>A p.R63Q Chr.2: 73902421-73902421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs922925882 Chr.2: 73902322-73902322 on Build GRCh38
rs200820569 Chr.2: 73902512-73902512 on Build GRCh38
rs372902508 Chr.2: 73902398-73902398 on Build GRCh38
rs769706469 Chr.2: 73902395-73902395 on Build GRCh38
rs750668174 Chr.2: 73902401-73902401 on Build GRCh38
rs372617230 Chr.2: 73902320-73902320 on Build GRCh38
rs745567878 Chr.2: 73902506-73902506 on Build GRCh38
rs752084477 Chr.2: 73902379-73902379 on Build GRCh38
rs864309491 Chr.2: 73902420-73902420 on Build GRCh38
rs1013083557 Chr.2: 73902533-73902533 on Build GRCh38
rs143614519 Chr.2: 73902479-73902479 on Build GRCh38
rs375008992 Chr.2: 73902523-73902523 on Build GRCh38
rs149132468 Chr.2: 73902455-73902455 on Build GRCh38
rs777727550 Chr.2: 73902459-73902459 on Build GRCh38
rs746915987 Chr.2: 73902469-73902469 on Build GRCh38
rs201925072 Chr.2: 73902336-73902336 on Build GRCh38
rs201402800 Chr.2: 73902428-73902428 on Build GRCh38
rs780136497 Chr.2: 73902450-73902450 on Build GRCh38
rs762594700 Chr.2: 73902521-73902521 on Build GRCh38
rs775787030 Chr.2: 73902505-73902505 on Build GRCh38
rs864309490 Chr.2: 73902367-73902367 on Build GRCh38
rs1726025 Chr.2: 73902347-73902347 on Build GRCh38
rs146195554 Chr.2: 73902318-73902318 on Build GRCh38
rs764750672 Chr.2: 73902353-73902353 on Build GRCh38
rs755027476 Chr.2: 73902458-73902458 on Build GRCh38
rs764809256 Chr.2: 73902325-73902325 on Build GRCh38
rs769524584 Chr.2: 73902510-73902510 on Build GRCh38
rs757710565 Chr.2: 73902387-73902387 on Build GRCh38
rs1653256 Chr.2: 73902537-73902537 on Build GRCh38

View Full Product Details