Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71065588-71065861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB2
Entrez Gene ID
9406
Gene Name
zinc finger RANBP2-type containing 2
Gene Aliases
ZIS, ZIS1, ZIS2, ZNF265
UniGene
Hs.194718
Gene Chromosome Location
Chr.1: 71063291-71081289 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Gene Symbol
ZRANB2-AS1
Entrez Gene ID
100132618
Gene Name
ZRANB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.720513
Gene Chromosome Location
Chr.1: 71046506-71067184 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1602659 c.934G>A p.V312M Chr.1: 71065748-71065748 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113219385 Chr.1: 71065799-71065799 on Build GRCh38
rs561885582 Chr.1: 71065705-71065705 on Build GRCh38
rs764254908 Chr.1: 71065744-71065744 on Build GRCh38
rs761334875 Chr.1: 71065757-71065758 on Build GRCh38
rs1008589279 Chr.1: 71065630-71065630 on Build GRCh38
rs778977071 Chr.1: 71065679-71065679 on Build GRCh38
rs371473086 Chr.1: 71065734-71065734 on Build GRCh38
rs771613490 Chr.1: 71065688-71065688 on Build GRCh38
rs779215123 Chr.1: 71065805-71065805 on Build GRCh38
rs747676859 Chr.1: 71065684-71065684 on Build GRCh38
rs889934751 Chr.1: 71065634-71065634 on Build GRCh38
rs374347376 Chr.1: 71065721-71065721 on Build GRCh38
rs752690355 Chr.1: 71065793-71065793 on Build GRCh38
rs770353425 Chr.1: 71065718-71065718 on Build GRCh38
rs762135525 Chr.1: 71065757-71065757 on Build GRCh38
rs974989630 Chr.1: 71065700-71065700 on Build GRCh38
rs574331789 Chr.1: 71065670-71065670 on Build GRCh38
rs754671148 Chr.1: 71065781-71065781 on Build GRCh38
rs541906223 Chr.1: 71065662-71065662 on Build GRCh38
rs934078028 Chr.1: 71065622-71065622 on Build GRCh38
rs529082358 Chr.1: 71065652-71065652 on Build GRCh38
rs774253148 Chr.1: 71065755-71065755 on Build GRCh38
rs776016247 Chr.1: 71065687-71065688 on Build GRCh38
rs777101445 Chr.1: 71065693-71065693 on Build GRCh38
rs753740436 Chr.1: 71065769-71065769 on Build GRCh38
rs964574432 Chr.1: 71065681-71065681 on Build GRCh38
rs370185269 Chr.1: 71065714-71065714 on Build GRCh38
rs1011973234 Chr.1: 71065809-71065809 on Build GRCh38
rs746708592 Chr.1: 71065698-71065698 on Build GRCh38
rs767824299 Chr.1: 71065758-71065758 on Build GRCh38
rs902761608 Chr.1: 71065732-71065732 on Build GRCh38
rs770462528 Chr.1: 71065701-71065701 on Build GRCh38
rs758231234 Chr.1: 71065798-71065798 on Build GRCh38
rs1019144172 Chr.1: 71065648-71065648 on Build GRCh38
rs765055492 Chr.1: 71065792-71065792 on Build GRCh38

View Full Product Details