Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112142048-112142321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
UniGene
Hs.5148
Gene Chromosome Location
Chr.12: 112125545-112153604 on Build GRCh38
Transcripts
NM_001143906.1 NM_006700.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1358823 c.796G>A p.V266I Chr.12: 112142241-112142241 on Build GRCh38
430351 c.838A>T p.S280C Chr.12: 112142283-112142283 on Build GRCh38
5374222 c.690C>T p.P230P Chr.12: 112142135-112142135 on Build GRCh38
270381 c.805G>A p.A269T Chr.12: 112142250-112142250 on Build GRCh38
6307924 c.700G>T p.G234C Chr.12: 112142145-112142145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765779444 Chr.12: 112142292-112142292 on Build GRCh38
rs762380014 Chr.12: 112142217-112142217 on Build GRCh38
rs747939315 Chr.12: 112142274-112142274 on Build GRCh38
rs61743643 Chr.12: 112142116-112142116 on Build GRCh38
rs115306073 Chr.12: 112142253-112142253 on Build GRCh38
rs769711871 Chr.12: 112142280-112142280 on Build GRCh38
rs140094553 Chr.12: 112142237-112142237 on Build GRCh38
rs771616039 Chr.12: 112142147-112142147 on Build GRCh38
rs542420544 Chr.12: 112142118-112142118 on Build GRCh38
rs61740854 Chr.12: 112142141-112142141 on Build GRCh38
rs560633197 Chr.12: 112142245-112142245 on Build GRCh38
rs779293514 Chr.12: 112142244-112142244 on Build GRCh38
rs1026034840 Chr.12: 112142144-112142144 on Build GRCh38
rs138174173 Chr.12: 112142194-112142194 on Build GRCh38
rs200342910 Chr.12: 112142182-112142182 on Build GRCh38
rs114577048 Chr.12: 112142268-112142268 on Build GRCh38
rs773048523 Chr.12: 112142283-112142283 on Build GRCh38
rs77125209 Chr.12: 112142213-112142213 on Build GRCh38
rs756571466 Chr.12: 112142134-112142134 on Build GRCh38
rs544336176 Chr.12: 112142210-112142210 on Build GRCh38
rs781535945 Chr.12: 112142252-112142252 on Build GRCh38
rs146374447 Chr.12: 112142231-112142231 on Build GRCh38
rs754314059 Chr.12: 112142241-112142241 on Build GRCh38
rs867096145 Chr.12: 112142133-112142133 on Build GRCh38
rs778388539 Chr.12: 112142135-112142135 on Build GRCh38
rs562893780 Chr.12: 112142212-112142212 on Build GRCh38
rs776401752 Chr.12: 112142270-112142270 on Build GRCh38
rs371156907 Chr.12: 112142180-112142180 on Build GRCh38
rs763968969 Chr.12: 112142078-112142078 on Build GRCh38
rs764778877 Chr.12: 112142104-112142104 on Build GRCh38
rs761777981 Chr.12: 112142098-112142098 on Build GRCh38
rs753804595 Chr.12: 112142089-112142089 on Build GRCh38
rs556936551 Chr.12: 112142096-112142096 on Build GRCh38
rs7970397 Chr.12: 112142267-112142267 on Build GRCh38
rs373642082 Chr.12: 112142085-112142086 on Build GRCh38
rs749955345 Chr.12: 112142110-112142110 on Build GRCh38
rs762488979 Chr.12: 112142291-112142291 on Build GRCh38
rs754722874 Chr.12: 112142240-112142240 on Build GRCh38
rs746004265 Chr.12: 112142160-112142160 on Build GRCh38
rs753050200 Chr.12: 112142121-112142121 on Build GRCh38
rs774057753 Chr.12: 112142295-112142295 on Build GRCh38
rs749878983 Chr.12: 112142216-112142216 on Build GRCh38
rs779037815 Chr.12: 112142155-112142155 on Build GRCh38
rs1012118598 Chr.12: 112142284-112142284 on Build GRCh38
rs768408627 Chr.12: 112142187-112142187 on Build GRCh38

View Full Product Details