Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4357875-4358148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO7
Entrez Gene ID
79585
Gene Name
coronin 7
Gene Aliases
0610011B16Rik, CRN7, POD1
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4354542-4416961 on Build GRCh38
Transcripts
NM_001201472.1 NM_001201473.1 NM_024535.4 NM_001201479.1

Gene Symbol
CORO7-PAM16
Entrez Gene ID
100529144
Gene Name
CORO7-PAM16 readthrough
Gene Aliases
-
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4340251-4416961 on Build GRCh38
Transcripts
NM_001201472.1 NM_001201473.1 NM_024535.4 NM_001201479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6487010 c.2480T>C p.V827A Chr.16: 4358081-4358081 on Build GRCh38
5850916 c.2494G>A p.A832T Chr.16: 4358067-4358067 on Build GRCh38
7269126 c.2464T>C p.F822L Chr.16: 4358097-4358097 on Build GRCh38
6634276 c.2555C>T p.P852L Chr.16: 4358006-4358006 on Build GRCh38
3509617 c.2503T>A p.W835R Chr.16: 4358058-4358058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553598882 Chr.16: 4358042-4358042 on Build GRCh38
rs915582010 Chr.16: 4357913-4357913 on Build GRCh38
rs778023615 Chr.16: 4357998-4357998 on Build GRCh38
rs745478194 Chr.16: 4358038-4358038 on Build GRCh38
rs202129042 Chr.16: 4357935-4357935 on Build GRCh38
rs753208669 Chr.16: 4358043-4358043 on Build GRCh38
rs368671302 Chr.16: 4358119-4358119 on Build GRCh38
rs762879819 Chr.16: 4358085-4358085 on Build GRCh38
rs777343569 Chr.16: 4357911-4357911 on Build GRCh38
rs140184459 Chr.16: 4358069-4358069 on Build GRCh38
rs937084996 Chr.16: 4357901-4357901 on Build GRCh38
rs748084320 Chr.16: 4358098-4358098 on Build GRCh38
rs769589131 Chr.16: 4358026-4358026 on Build GRCh38
rs769595679 Chr.16: 4358049-4358049 on Build GRCh38
rs746917422 Chr.16: 4357932-4357932 on Build GRCh38
rs776405953 Chr.16: 4357943-4357943 on Build GRCh38
rs764955995 Chr.16: 4357948-4357948 on Build GRCh38
rs34507965 Chr.16: 4357972-4357973 on Build GRCh38
rs763208781 Chr.16: 4358034-4358034 on Build GRCh38
rs779594104 Chr.16: 4358005-4358005 on Build GRCh38
rs764489771 Chr.16: 4358054-4358054 on Build GRCh38
rs929449517 Chr.16: 4357993-4357993 on Build GRCh38
rs758055933 Chr.16: 4358068-4358068 on Build GRCh38
rs750054519 Chr.16: 4358000-4358000 on Build GRCh38
rs763483206 Chr.16: 4357928-4357928 on Build GRCh38
rs774891907 Chr.16: 4357929-4357929 on Build GRCh38
rs566858497 Chr.16: 4358044-4358044 on Build GRCh38
rs370883825 Chr.16: 4358033-4358033 on Build GRCh38
rs777607959 Chr.16: 4358122-4358122 on Build GRCh38
rs751226091 Chr.16: 4358073-4358073 on Build GRCh38
rs568562521 Chr.16: 4357937-4357937 on Build GRCh38
rs201607278 Chr.16: 4358037-4358037 on Build GRCh38
rs749100014 Chr.16: 4358126-4358126 on Build GRCh38
rs760111111 Chr.16: 4357966-4357966 on Build GRCh38
rs191380280 Chr.16: 4357973-4357973 on Build GRCh38
rs753263864 Chr.16: 4357983-4357983 on Build GRCh38
rs200635962 Chr.16: 4357961-4357961 on Build GRCh38
rs144324307 Chr.16: 4358062-4358062 on Build GRCh38
rs763118244 Chr.16: 4357958-4357958 on Build GRCh38
rs549223912 Chr.16: 4357912-4357912 on Build GRCh38
rs756446929 Chr.16: 4357992-4357992 on Build GRCh38
rs773437672 Chr.16: 4357952-4357952 on Build GRCh38
rs900514322 Chr.16: 4358061-4358061 on Build GRCh38
rs139084024 Chr.16: 4358001-4358001 on Build GRCh38
rs112338816 Chr.16: 4357900-4357900 on Build GRCh38
rs780287127 Chr.16: 4357937-4357937 on Build GRCh38
rs761530558 Chr.16: 4357946-4357946 on Build GRCh38
rs969746752 Chr.16: 4357936-4357936 on Build GRCh38
rs776114367 Chr.16: 4358020-4358020 on Build GRCh38
rs766454949 Chr.16: 4357960-4357960 on Build GRCh38
rs555588403 Chr.16: 4358083-4358083 on Build GRCh38
rs149619313 Chr.16: 4358011-4358011 on Build GRCh38

View Full Product Details