Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4956546-4956817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENO3
Entrez Gene ID
2027
Gene Name
enolase 3
Gene Aliases
GSD13, MSE
UniGene
Hs.224171
Gene Chromosome Location
Chr.17: 4949182-4957131 on Build GRCh38
Transcripts
NM_001193503.1 NM_001976.4 NM_053013.3 XM_005256521.2 XM_011523729.1 XM_017024346.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4579837 c.1164C>G p.L388L Chr.17: 4956669-4956669 on Build GRCh38
1384481 c.1094delG p.V367fs*1 Chr.17: 4956599-4956599 on Build GRCh38
4680988 c.1114C>T p.R372C Chr.17: 4956619-4956619 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs990512242 Chr.17: 4956791-4956791 on Build GRCh38
rs773176585 Chr.17: 4956569-4956569 on Build GRCh38
rs769879612 Chr.17: 4956619-4956619 on Build GRCh38
rs758682068 Chr.17: 4956687-4956687 on Build GRCh38
rs751261485 Chr.17: 4956576-4956576 on Build GRCh38
rs199919172 Chr.17: 4956724-4956724 on Build GRCh38
rs770822790 Chr.17: 4956628-4956628 on Build GRCh38
rs745468546 Chr.17: 4956782-4956782 on Build GRCh38
rs748062955 Chr.17: 4956612-4956612 on Build GRCh38
rs754410640 Chr.17: 4956669-4956669 on Build GRCh38
rs991804133 Chr.17: 4956697-4956697 on Build GRCh38
rs924623854 Chr.17: 4956755-4956755 on Build GRCh38
rs756615985 Chr.17: 4956597-4956597 on Build GRCh38
rs367559124 Chr.17: 4956780-4956780 on Build GRCh38
rs121918404 Chr.17: 4956626-4956626 on Build GRCh38
rs753189024 Chr.17: 4956662-4956663 on Build GRCh38
rs771868807 Chr.17: 4956597-4956598 on Build GRCh38
rs773860245 Chr.17: 4956632-4956632 on Build GRCh38
rs780289212 Chr.17: 4956611-4956611 on Build GRCh38
rs976110845 Chr.17: 4956748-4956748 on Build GRCh38
rs750547165 Chr.17: 4956684-4956684 on Build GRCh38
rs778070292 Chr.17: 4956598-4956598 on Build GRCh38
rs757550331 Chr.17: 4956671-4956671 on Build GRCh38
rs780236325 Chr.17: 4956688-4956688 on Build GRCh38
rs778998819 Chr.17: 4956607-4956607 on Build GRCh38
rs758962137 Chr.17: 4956580-4956580 on Build GRCh38
rs770572476 Chr.17: 4956727-4956727 on Build GRCh38
rs916301975 Chr.17: 4956720-4956720 on Build GRCh38
rs781779699 Chr.17: 4956703-4956703 on Build GRCh38
rs961001512 Chr.17: 4956599-4956599 on Build GRCh38
rs773328325 Chr.17: 4956620-4956620 on Build GRCh38
rs765650202 Chr.17: 4956674-4956674 on Build GRCh38
rs757679931 Chr.17: 4956605-4956605 on Build GRCh38
rs766870438 Chr.17: 4956638-4956638 on Build GRCh38
rs371648627 Chr.17: 4956572-4956572 on Build GRCh38
rs761121808 Chr.17: 4956665-4956665 on Build GRCh38
rs768012807 Chr.17: 4956658-4956660 on Build GRCh38
rs952351975 Chr.17: 4956642-4956642 on Build GRCh38
rs111926990 Chr.17: 4956689-4956689 on Build GRCh38
rs758355479 Chr.17: 4956610-4956610 on Build GRCh38
rs762649721 Chr.17: 4956571-4956571 on Build GRCh38
rs755106167 Chr.17: 4956659-4956659 on Build GRCh38
rs759238253 Chr.17: 4956637-4956637 on Build GRCh38
rs550460218 Chr.17: 4956600-4956600 on Build GRCh38
rs764554186 Chr.17: 4956667-4956667 on Build GRCh38
rs370533034 Chr.17: 4956691-4956691 on Build GRCh38
rs1043861567 Chr.17: 4956723-4956723 on Build GRCh38
rs761089932 Chr.17: 4956688-4956689 on Build GRCh38
rs533687145 Chr.17: 4956594-4956594 on Build GRCh38
rs447251 Chr.17: 4956721-4956721 on Build GRCh38
rs767111011 Chr.17: 4956589-4956589 on Build GRCh38
rs951950275 Chr.17: 4956658-4956658 on Build GRCh38
rs369680560 Chr.17: 4956745-4956745 on Build GRCh38
rs775011353 Chr.17: 4956657-4956657 on Build GRCh38
rs746160926 Chr.17: 4956608-4956608 on Build GRCh38
rs1027911959 Chr.17: 4956652-4956652 on Build GRCh38

View Full Product Details