Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 64971502-64971771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH11
Entrez Gene ID
1009
Gene Name
cadherin 11
Gene Aliases
CAD11, CDHOB, OB, OSF-4
UniGene
Hs.116471
Gene Chromosome Location
Chr.16: 64943753-65126112 on Build GRCh38
Transcripts
NM_001308392.1 NM_001797.3 XM_005255761.3 XM_005255762.2 XM_005255763.2 XM_011522803.2 XM_017022830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4782979 c.1642G>T p.D548Y Chr.16: 64971579-64971579 on Build GRCh38
4528670 c.1555G>A p.D519N Chr.16: 64971666-64971666 on Build GRCh38
5748970 c.1561A>G p.T521A Chr.16: 64971660-64971660 on Build GRCh38
5451402 c.1565C>T p.A522V Chr.16: 64971656-64971656 on Build GRCh38
2993405 c.1595C>A p.P532H Chr.16: 64971626-64971626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760526230 Chr.16: 64971565-64971565 on Build GRCh38
rs537542997 Chr.16: 64971738-64971738 on Build GRCh38
rs759138385 Chr.16: 64971584-64971584 on Build GRCh38
rs759584951 Chr.16: 64971668-64971668 on Build GRCh38
rs370410162 Chr.16: 64971558-64971558 on Build GRCh38
rs772815993 Chr.16: 64971657-64971657 on Build GRCh38
rs764349192 Chr.16: 64971625-64971625 on Build GRCh38
rs570752275 Chr.16: 64971567-64971567 on Build GRCh38
rs750909050 Chr.16: 64971741-64971741 on Build GRCh38
rs758115170 Chr.16: 64971614-64971614 on Build GRCh38
rs756157853 Chr.16: 64971647-64971647 on Build GRCh38
rs758715267 Chr.16: 64971532-64971532 on Build GRCh38
rs201159992 Chr.16: 64971587-64971587 on Build GRCh38
rs780231674 Chr.16: 64971648-64971648 on Build GRCh38
rs768990129 Chr.16: 64971654-64971654 on Build GRCh38
rs745910479 Chr.16: 64971644-64971644 on Build GRCh38
rs373610980 Chr.16: 64971581-64971581 on Build GRCh38
rs751824771 Chr.16: 64971709-64971709 on Build GRCh38
rs535764382 Chr.16: 64971673-64971673 on Build GRCh38
rs369888999 Chr.16: 64971693-64971693 on Build GRCh38
rs775120047 Chr.16: 64971686-64971686 on Build GRCh38
rs202063749 Chr.16: 64971703-64971703 on Build GRCh38
rs200404897 Chr.16: 64971658-64971658 on Build GRCh38
rs769958307 Chr.16: 64971552-64971552 on Build GRCh38
rs762199530 Chr.16: 64971730-64971730 on Build GRCh38
rs752258359 Chr.16: 64971605-64971605 on Build GRCh38
rs757815099 Chr.16: 64971538-64971538 on Build GRCh38
rs200031328 Chr.16: 64971680-64971680 on Build GRCh38
rs199542657 Chr.16: 64971541-64971541 on Build GRCh38
rs749459591 Chr.16: 64971651-64971651 on Build GRCh38
rs923823612 Chr.16: 64971566-64971566 on Build GRCh38
rs376455105 Chr.16: 64971537-64971537 on Build GRCh38
rs773118377 Chr.16: 64971560-64971560 on Build GRCh38
rs12597529 Chr.16: 64971559-64971559 on Build GRCh38
rs757706888 Chr.16: 64971637-64971637 on Build GRCh38
rs776739314 Chr.16: 64971662-64971662 on Build GRCh38
rs781683134 Chr.16: 64971642-64971642 on Build GRCh38
rs781186581 Chr.16: 64971540-64971540 on Build GRCh38
rs879128241 Chr.16: 64971682-64971682 on Build GRCh38
rs377418741 Chr.16: 64971551-64971551 on Build GRCh38
rs566774546 Chr.16: 64971659-64971659 on Build GRCh38
rs751962998 Chr.16: 64971630-64971630 on Build GRCh38
rs868653167 Chr.16: 64971621-64971621 on Build GRCh38
rs1038101855 Chr.16: 64971715-64971715 on Build GRCh38
rs756673090 Chr.16: 64971743-64971743 on Build GRCh38

View Full Product Details