Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 841847-842118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUN1
Entrez Gene ID
23353
Gene Name
Sad1 and UNC84 domain containing 1
Gene Aliases
UNC84A
UniGene
Hs.438072
Gene Chromosome Location
Chr.7: 815557-874920 on Build GRCh38
Transcripts
NM_001130965.2 NM_001171944.1 NM_001171945.1 NM_001171946.1 NM_025154.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4998810 c.352C>T p.H118Y Chr.7: 842031-842031 on Build GRCh38
4981113 c.296A>G p.K99R Chr.7: 841975-841975 on Build GRCh38
6872619 c.499G>A p.V167M Chr.7: 842115-842115 on Build GRCh38
3642037 c.405G>A p.L135L Chr.7: 842084-842084 on Build GRCh38
6571921 c.198C>T p.V66V Chr.7: 842027-842027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753262058 Chr.7: 842040-842040 on Build GRCh38
rs771878466 Chr.7: 842088-842088 on Build GRCh38
rs781471011 Chr.7: 842068-842068 on Build GRCh38
rs767236789 Chr.7: 842033-842033 on Build GRCh38
rs571168917 Chr.7: 841912-841912 on Build GRCh38
rs747459970 Chr.7: 842010-842010 on Build GRCh38
rs768195590 Chr.7: 841909-841909 on Build GRCh38
rs1023474662 Chr.7: 842032-842032 on Build GRCh38
rs903549010 Chr.7: 841970-841970 on Build GRCh38
rs778427452 Chr.7: 842005-842005 on Build GRCh38
rs775583098 Chr.7: 842083-842083 on Build GRCh38
rs767433187 Chr.7: 841952-841952 on Build GRCh38
rs756278052 Chr.7: 841929-841929 on Build GRCh38
rs912678292 Chr.7: 842072-842072 on Build GRCh38
rs747866078 Chr.7: 841939-841939 on Build GRCh38
rs921546955 Chr.7: 841937-841937 on Build GRCh38
rs199666815 Chr.7: 842037-842037 on Build GRCh38
rs541487561 Chr.7: 842041-842041 on Build GRCh38
rs771160146 Chr.7: 842013-842013 on Build GRCh38
rs759842974 Chr.7: 842015-842015 on Build GRCh38
rs950522872 Chr.7: 841892-841892 on Build GRCh38
rs562530676 Chr.7: 841875-841875 on Build GRCh38
rs367835809 Chr.7: 842077-842077 on Build GRCh38
rs78842948 Chr.7: 841957-841957 on Build GRCh38
rs527315917 Chr.7: 841929-841929 on Build GRCh38
rs999120053 Chr.7: 841989-841989 on Build GRCh38
rs772702463 Chr.7: 841999-841999 on Build GRCh38
rs772436237 Chr.7: 841901-841901 on Build GRCh38
rs761859949 Chr.7: 841941-841941 on Build GRCh38
rs538774385 Chr.7: 842024-842024 on Build GRCh38
rs567829498 Chr.7: 841907-841907 on Build GRCh38
rs748728198 Chr.7: 841897-841897 on Build GRCh38
rs376335038 Chr.7: 841998-841998 on Build GRCh38
rs745922989 Chr.7: 842070-842070 on Build GRCh38
rs912635938 Chr.7: 841886-841886 on Build GRCh38
rs199937447 Chr.7: 842065-842065 on Build GRCh38
rs779105133 Chr.7: 841895-841895 on Build GRCh38
rs370786270 Chr.7: 842036-842036 on Build GRCh38
rs751282249 Chr.7: 841939-841939 on Build GRCh38
rs763981938 Chr.7: 841932-841932 on Build GRCh38
rs1025688681 Chr.7: 841890-841890 on Build GRCh38
rs535135658 Chr.7: 841928-841929 on Build GRCh38
rs773479778 Chr.7: 841904-841904 on Build GRCh38
rs778231328 Chr.7: 842054-842054 on Build GRCh38
rs771066327 Chr.7: 841917-841917 on Build GRCh38
rs371837294 Chr.7: 842061-842061 on Build GRCh38
rs536026654 Chr.7: 841968-841968 on Build GRCh38
rs551327704 Chr.7: 841885-841885 on Build GRCh38
rs777003860 Chr.7: 841926-841926 on Build GRCh38
rs909068458 Chr.7: 841908-841908 on Build GRCh38
rs6461378 Chr.7: 842031-842031 on Build GRCh38
rs183872262 Chr.7: 842014-842014 on Build GRCh38
rs112598200 Chr.7: 841960-841960 on Build GRCh38
rs575219579 Chr.7: 842087-842087 on Build GRCh38
rs555948337 Chr.7: 842012-842012 on Build GRCh38
rs771856676 Chr.7: 841987-841987 on Build GRCh38
rs374613091 Chr.7: 842025-842025 on Build GRCh38
rs754552795 Chr.7: 841977-841977 on Build GRCh38
rs41273066 Chr.7: 842045-842045 on Build GRCh38
rs531311600 Chr.7: 841884-841884 on Build GRCh38
rs752326487 Chr.7: 842049-842049 on Build GRCh38
rs769303158 Chr.7: 841980-841980 on Build GRCh38
rs758701308 Chr.7: 842051-842051 on Build GRCh38
rs941926809 Chr.7: 841916-841916 on Build GRCh38
rs779380234 Chr.7: 841990-841990 on Build GRCh38

View Full Product Details