Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38377641-38377906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs763427872 Chr.17: 38377749-38377749 on Build GRCh38
rs140480789 Chr.17: 38377754-38377754 on Build GRCh38
rs746955104 Chr.17: 38377839-38377839 on Build GRCh38
rs764623857 Chr.17: 38377752-38377752 on Build GRCh38
rs760589332 Chr.17: 38377673-38377673 on Build GRCh38
rs776516873 Chr.17: 38377855-38377855 on Build GRCh38
rs762516127 Chr.17: 38377872-38377872 on Build GRCh38
rs758382136 Chr.17: 38377812-38377812 on Build GRCh38
rs779952709 Chr.17: 38377788-38377788 on Build GRCh38
rs770562468 Chr.17: 38377851-38377851 on Build GRCh38
rs1030282493 Chr.17: 38377691-38377691 on Build GRCh38
rs749431653 Chr.17: 38377811-38377811 on Build GRCh38
rs769535716 Chr.17: 38377867-38377867 on Build GRCh38
rs774949800 Chr.17: 38377871-38377871 on Build GRCh38
rs995568815 Chr.17: 38377833-38377833 on Build GRCh38
rs554906158 Chr.17: 38377823-38377823 on Build GRCh38
rs759227951 Chr.17: 38377694-38377694 on Build GRCh38
rs773667160 Chr.17: 38377678-38377679 on Build GRCh38
rs770601889 Chr.17: 38377687-38377687 on Build GRCh38
rs750545104 Chr.17: 38377781-38377781 on Build GRCh38
rs752226648 Chr.17: 38377742-38377742 on Build GRCh38
rs764570283 Chr.17: 38377878-38377878 on Build GRCh38
rs768415531 Chr.17: 38377866-38377866 on Build GRCh38
rs751915429 Chr.17: 38377753-38377753 on Build GRCh38
rs763040902 Chr.17: 38377692-38377692 on Build GRCh38
rs755069583 Chr.17: 38377717-38377717 on Build GRCh38
rs764608662 Chr.17: 38377727-38377727 on Build GRCh38
rs781630898 Chr.17: 38377780-38377780 on Build GRCh38
rs147043285 Chr.17: 38377787-38377787 on Build GRCh38
rs776086094 Chr.17: 38377688-38377688 on Build GRCh38

View Full Product Details