Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42250412-42250671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERF
Entrez Gene ID
2077
Gene Name
ETS2 repressor factor
Gene Aliases
CRS4, PE-2, PE2
UniGene
Hs.655969
Gene Chromosome Location
Chr.19: 42247561-42255164 on Build GRCh38
Transcripts
NM_001301035.1 NM_001308402.1 NM_001312656.1 NM_006494.3 XM_017026468.1 XM_017026469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855936 c.68G>A p.G23D Chr.19: 42250520-42250520 on Build GRCh38
1290961 c.23-1G>T p.? Chr.19: 42250566-42250566 on Build GRCh38
3534668 c.96T>G p.F32L Chr.19: 42250492-42250492 on Build GRCh38
5542894 c.102G>A p.L34L Chr.19: 42250486-42250486 on Build GRCh38
5487423 c.157G>C p.G53R Chr.19: 42250431-42250431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538509042 Chr.19: 42250528-42250528 on Build GRCh38
rs898918186 Chr.19: 42250545-42250545 on Build GRCh38
rs1037421856 Chr.19: 42250535-42250535 on Build GRCh38
rs748401659 Chr.19: 42250584-42250584 on Build GRCh38
rs763079808 Chr.19: 42250609-42250609 on Build GRCh38
rs864321680 Chr.19: 42250567-42250567 on Build GRCh38
rs545519539 Chr.19: 42250594-42250594 on Build GRCh38
rs753578618 Chr.19: 42250510-42250510 on Build GRCh38
rs199685654 Chr.19: 42250574-42250574 on Build GRCh38
rs769938254 Chr.19: 42250588-42250588 on Build GRCh38
rs914128913 Chr.19: 42250455-42250455 on Build GRCh38
rs767745561 Chr.19: 42250504-42250504 on Build GRCh38
rs769908132 Chr.19: 42250451-42250451 on Build GRCh38
rs761095300 Chr.19: 42250568-42250569 on Build GRCh38
rs371076249 Chr.19: 42250552-42250552 on Build GRCh38
rs1019339310 Chr.19: 42250582-42250582 on Build GRCh38
rs192294053 Chr.19: 42250456-42250456 on Build GRCh38
rs763456687 Chr.19: 42250475-42250475 on Build GRCh38
rs371399184 Chr.19: 42250573-42250573 on Build GRCh38
rs375067175 Chr.19: 42250595-42250595 on Build GRCh38
rs150347249 Chr.19: 42250438-42250438 on Build GRCh38
rs375582175 Chr.19: 42250555-42250555 on Build GRCh38
rs200969503 Chr.19: 42250564-42250564 on Build GRCh38
rs746973324 Chr.19: 42250519-42250519 on Build GRCh38
rs778594748 Chr.19: 42250543-42250543 on Build GRCh38
rs111300456 Chr.19: 42250481-42250481 on Build GRCh38
rs766851829 Chr.19: 42250486-42250486 on Build GRCh38
rs746916094 Chr.19: 42250569-42250569 on Build GRCh38
rs569171834 Chr.19: 42250558-42250558 on Build GRCh38
rs751996940 Chr.19: 42250489-42250489 on Build GRCh38
rs1051849829 Chr.19: 42250480-42250480 on Build GRCh38
rs760016604 Chr.19: 42250495-42250495 on Build GRCh38
rs776450665 Chr.19: 42250631-42250631 on Build GRCh38

View Full Product Details