Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98521504-98521731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDND1
Entrez Gene ID
56650
Gene Name
claudin domain containing 1
Gene Aliases
C3orf4, GENX-3745
UniGene
Hs.531371
Gene Chromosome Location
Chr.3: 98515473-98523066 on Build GRCh38
Transcripts
NM_001040181.1 NM_001040182.1 NM_001040183.1 NM_001040199.1 NM_001040200.1 NM_019895.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982278 c.10G>A p.D4N Chr.3: 98521694-98521694 on Build GRCh38
7410672 c.28A>T p.T10S Chr.3: 98521676-98521676 on Build GRCh38
7094961 c.51+1G>A p.? Chr.3: 98521652-98521652 on Build GRCh38
6041994 c.19G>C p.E7Q Chr.3: 98521685-98521685 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376270288 Chr.3: 98521635-98521635 on Build GRCh38
rs371059754 Chr.3: 98521626-98521626 on Build GRCh38
rs753880425 Chr.3: 98521656-98521656 on Build GRCh38
rs112805874 Chr.3: 98521698-98521698 on Build GRCh38
rs189730306 Chr.3: 98521552-98521552 on Build GRCh38
rs992089433 Chr.3: 98521532-98521532 on Build GRCh38
rs367625149 Chr.3: 98521653-98521653 on Build GRCh38
rs766490832 Chr.3: 98521580-98521580 on Build GRCh38
rs11553640 Chr.3: 98521665-98521665 on Build GRCh38
rs745369156 Chr.3: 98521685-98521685 on Build GRCh38
rs924389722 Chr.3: 98521550-98521550 on Build GRCh38
rs1054306146 Chr.3: 98521569-98521569 on Build GRCh38
rs151010606 Chr.3: 98521657-98521657 on Build GRCh38
rs917706472 Chr.3: 98521554-98521554 on Build GRCh38
rs763283566 Chr.3: 98521610-98521610 on Build GRCh38
rs996790875 Chr.3: 98521607-98521607 on Build GRCh38
rs780290258 Chr.3: 98521670-98521671 on Build GRCh38
rs200126709 Chr.3: 98521684-98521684 on Build GRCh38
rs755018640 Chr.3: 98521639-98521639 on Build GRCh38
rs867636423 Chr.3: 98521693-98521693 on Build GRCh38
rs764420328 Chr.3: 98521619-98521619 on Build GRCh38
rs950612986 Chr.3: 98521594-98521594 on Build GRCh38
rs756797664 Chr.3: 98521679-98521679 on Build GRCh38
rs746582990 Chr.3: 98521670-98521670 on Build GRCh38
rs750509365 Chr.3: 98521637-98521637 on Build GRCh38
rs1041890438 Chr.3: 98521543-98521543 on Build GRCh38

View Full Product Details