Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46623681-46623954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIPT
Entrez Gene ID
9419
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Gene Aliases
HSPC139, SSMDF
UniGene
Hs.133998
Gene Chromosome Location
Chr.2: 46617172-46630176 on Build GRCh38
Transcripts
NM_014171.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
477451 c.158A>G p.N53S Chr.2: 46623784-46623784 on Build GRCh38
4399627 c.146C>T p.P49L Chr.2: 46623772-46623772 on Build GRCh38
6429033 c.170C>G p.T57S Chr.2: 46623796-46623796 on Build GRCh38
3426482 c.156G>T p.K52N Chr.2: 46623782-46623782 on Build GRCh38
1021196 c.189T>G p.S63R Chr.2: 46623815-46623815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759308336 Chr.2: 46623735-46623735 on Build GRCh38
rs780701459 Chr.2: 46623899-46623899 on Build GRCh38
rs374987044 Chr.2: 46623872-46623872 on Build GRCh38
rs766689475 Chr.2: 46623876-46623876 on Build GRCh38
rs371586247 Chr.2: 46623843-46623843 on Build GRCh38
rs375390924 Chr.2: 46623704-46623704 on Build GRCh38
rs556367308 Chr.2: 46623760-46623760 on Build GRCh38
rs183532442 Chr.2: 46623715-46623715 on Build GRCh38
rs192909311 Chr.2: 46623874-46623874 on Build GRCh38
rs767688021 Chr.2: 46623781-46623781 on Build GRCh38
rs776324669 Chr.2: 46623734-46623734 on Build GRCh38
rs745536966 Chr.2: 46623905-46623905 on Build GRCh38
rs762934398 Chr.2: 46623755-46623755 on Build GRCh38
rs587779348 Chr.2: 46623767-46623767 on Build GRCh38
rs769564437 Chr.2: 46623743-46623743 on Build GRCh38
rs771586131 Chr.2: 46623721-46623721 on Build GRCh38
rs746696706 Chr.2: 46623728-46623728 on Build GRCh38
rs750649993 Chr.2: 46623782-46623782 on Build GRCh38
rs187870794 Chr.2: 46623717-46623717 on Build GRCh38
rs756731005 Chr.2: 46623898-46623898 on Build GRCh38
rs367572446 Chr.2: 46623802-46623802 on Build GRCh38
rs775301061 Chr.2: 46623749-46623749 on Build GRCh38
rs762807861 Chr.2: 46623839-46623839 on Build GRCh38
rs60085835 Chr.2: 46623702-46623702 on Build GRCh38
rs10194490 Chr.2: 46623886-46623886 on Build GRCh38
rs902964540 Chr.2: 46623926-46623926 on Build GRCh38
rs746571899 Chr.2: 46623811-46623811 on Build GRCh38
rs755440723 Chr.2: 46623800-46623800 on Build GRCh38
rs529659701 Chr.2: 46623757-46623757 on Build GRCh38
rs554039395 Chr.2: 46623847-46623847 on Build GRCh38
rs985754806 Chr.2: 46623859-46623859 on Build GRCh38
rs888777600 Chr.2: 46623909-46623909 on Build GRCh38
rs1021105685 Chr.2: 46623757-46623757 on Build GRCh38
rs780410895 Chr.2: 46623795-46623795 on Build GRCh38
rs370491931 Chr.2: 46623773-46623773 on Build GRCh38
rs577929077 Chr.2: 46623775-46623775 on Build GRCh38
rs201527688 Chr.2: 46623725-46623725 on Build GRCh38
rs60552651 Chr.2: 46623759-46623759 on Build GRCh38
rs779826892 Chr.2: 46623914-46623914 on Build GRCh38
rs140621301 Chr.2: 46623827-46623827 on Build GRCh38
rs375275632 Chr.2: 46623796-46623796 on Build GRCh38
rs770462964 Chr.2: 46623816-46623816 on Build GRCh38
rs896565175 Chr.2: 46623748-46623748 on Build GRCh38
rs919030540 Chr.2: 46623906-46623906 on Build GRCh38
rs376567583 Chr.2: 46623733-46623733 on Build GRCh38
rs767570879 Chr.2: 46623868-46623868 on Build GRCh38
rs755830661 Chr.2: 46623912-46623912 on Build GRCh38
rs775314824 Chr.2: 46623832-46623832 on Build GRCh38
rs200980926 Chr.2: 46623842-46623842 on Build GRCh38
rs80325258 Chr.2: 46623887-46623887 on Build GRCh38
rs745465203 Chr.2: 46623826-46623826 on Build GRCh38
rs377397776 Chr.2: 46623772-46623772 on Build GRCh38
rs749149408 Chr.2: 46623919-46623919 on Build GRCh38
rs11555851 Chr.2: 46623814-46623814 on Build GRCh38
rs375537241 Chr.2: 46623817-46623817 on Build GRCh38
rs1040130553 Chr.2: 46623925-46623925 on Build GRCh38
rs756443318 Chr.2: 46623794-46623794 on Build GRCh38

View Full Product Details