Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51367131-51367356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDND2
Entrez Gene ID
125875
Gene Name
claudin domain containing 2
Gene Aliases
-
UniGene
Hs.720536
Gene Chromosome Location
Chr.19: 51366750-51369255 on Build GRCh38
Transcripts
NM_152353.2 XM_011526425.2 XM_011526428.2 XM_017026244.1 XM_017026245.1 XM_017026246.1 XM_017026247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3537595 c.490C>T p.P164S Chr.19: 51367156-51367156 on Build GRCh38
6630071 c.431-3delC p.? Chr.19: 51367218-51367218 on Build GRCh38
5431758 c.431-1G>A p.? Chr.19: 51367216-51367216 on Build GRCh38
1631039 c.442C>T p.L148L Chr.19: 51367204-51367204 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780892260 Chr.19: 51367277-51367277 on Build GRCh38
rs199756062 Chr.19: 51367201-51367201 on Build GRCh38
rs376914909 Chr.19: 51367247-51367247 on Build GRCh38
rs777453479 Chr.19: 51367199-51367201 on Build GRCh38
rs529723160 Chr.19: 51367281-51367281 on Build GRCh38
rs777181361 Chr.19: 51367155-51367155 on Build GRCh38
rs765627231 Chr.19: 51367154-51367154 on Build GRCh38
rs751539630 Chr.19: 51367194-51367194 on Build GRCh38
rs142279589 Chr.19: 51367223-51367223 on Build GRCh38
rs745434055 Chr.19: 51367321-51367321 on Build GRCh38
rs141450057 Chr.19: 51367172-51367172 on Build GRCh38
rs755754582 Chr.19: 51367215-51367215 on Build GRCh38
rs748682043 Chr.19: 51367219-51367219 on Build GRCh38
rs781767827 Chr.19: 51367329-51367329 on Build GRCh38
rs185510103 Chr.19: 51367184-51367184 on Build GRCh38
rs766161552 Chr.19: 51367191-51367191 on Build GRCh38
rs773485101 Chr.19: 51367255-51367255 on Build GRCh38
rs756918706 Chr.19: 51367225-51367226 on Build GRCh38
rs1059344 Chr.19: 51367175-51367175 on Build GRCh38
rs751678679 Chr.19: 51367272-51367272 on Build GRCh38
rs753548685 Chr.19: 51367291-51367291 on Build GRCh38
rs950596693 Chr.19: 51367308-51367308 on Build GRCh38
rs778451424 Chr.19: 51367309-51367309 on Build GRCh38
rs762325155 Chr.19: 51367238-51367238 on Build GRCh38
rs759466006 Chr.19: 51367273-51367273 on Build GRCh38
rs199917528 Chr.19: 51367168-51367168 on Build GRCh38
rs769848269 Chr.19: 51367240-51367240 on Build GRCh38
rs763616424 Chr.19: 51367283-51367283 on Build GRCh38
rs748455692 Chr.19: 51367334-51367334 on Build GRCh38
rs767504860 Chr.19: 51367274-51367274 on Build GRCh38
rs752559823 Chr.19: 51367211-51367211 on Build GRCh38
rs190229451 Chr.19: 51367279-51367279 on Build GRCh38
rs987691494 Chr.19: 51367318-51367318 on Build GRCh38
rs769322408 Chr.19: 51367230-51367230 on Build GRCh38
rs934245622 Chr.19: 51367256-51367256 on Build GRCh38
rs548278507 Chr.19: 51367327-51367327 on Build GRCh38
rs756905229 Chr.19: 51367293-51367293 on Build GRCh38
rs565235431 Chr.19: 51367220-51367220 on Build GRCh38
rs543787993 Chr.19: 51367257-51367257 on Build GRCh38

View Full Product Details