Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 61871881-61872144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT4
Entrez Gene ID
10575
Gene Name
chaperonin containing TCP1 subunit 4
Gene Aliases
CCT-DELTA, Cctd, SRB
UniGene
Hs.421509
Gene Chromosome Location
Chr.2: 61868127-61888671 on Build GRCh38
Transcripts
NM_001256721.1 NM_006430.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022160 c.1449G>A p.R483R Chr.2: 61872124-61872124 on Build GRCh38
6996230 c.1486C>T p.R496* Chr.2: 61872087-61872087 on Build GRCh38
5749317 c.1462G>A p.E488K Chr.2: 61872111-61872111 on Build GRCh38
1408931 c.1468delA p.T490fs*30 Chr.2: 61872105-61872105 on Build GRCh38
4640451 c.1447C>T p.R483W Chr.2: 61872126-61872126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575785394 Chr.2: 61871996-61871996 on Build GRCh38
rs375740937 Chr.2: 61872052-61872052 on Build GRCh38
rs539807074 Chr.2: 61872085-61872085 on Build GRCh38
rs945194187 Chr.2: 61872028-61872028 on Build GRCh38
rs947036297 Chr.2: 61872013-61872013 on Build GRCh38
rs779010252 Chr.2: 61872055-61872055 on Build GRCh38
rs745657457 Chr.2: 61872066-61872066 on Build GRCh38
rs758610994 Chr.2: 61872057-61872057 on Build GRCh38
rs976976780 Chr.2: 61871909-61871909 on Build GRCh38
rs746530000 Chr.2: 61872046-61872049 on Build GRCh38
rs910230548 Chr.2: 61871959-61871959 on Build GRCh38
rs367617969 Chr.2: 61871938-61871938 on Build GRCh38
rs761185511 Chr.2: 61872044-61872044 on Build GRCh38
rs761512849 Chr.2: 61872109-61872109 on Build GRCh38
rs753872178 Chr.2: 61872051-61872051 on Build GRCh38
rs751611307 Chr.2: 61871960-61871960 on Build GRCh38
rs202089088 Chr.2: 61872042-61872042 on Build GRCh38
rs769137529 Chr.2: 61872067-61872067 on Build GRCh38
rs771635404 Chr.2: 61872115-61872115 on Build GRCh38
rs372593676 Chr.2: 61872046-61872046 on Build GRCh38
rs774070252 Chr.2: 61872108-61872108 on Build GRCh38
rs779663340 Chr.2: 61872039-61872043 on Build GRCh38
rs749287965 Chr.2: 61872099-61872099 on Build GRCh38
rs1002299577 Chr.2: 61872063-61872063 on Build GRCh38
rs933073637 Chr.2: 61871953-61871953 on Build GRCh38
rs578243728 Chr.2: 61871942-61871942 on Build GRCh38
rs368273591 Chr.2: 61871984-61871984 on Build GRCh38
rs938468176 Chr.2: 61871932-61871932 on Build GRCh38
rs3828276 Chr.2: 61872021-61872021 on Build GRCh38
rs755028246 Chr.2: 61871965-61871965 on Build GRCh38
rs572611884 Chr.2: 61872059-61872059 on Build GRCh38
rs542900203 Chr.2: 61872000-61872000 on Build GRCh38
rs780164717 Chr.2: 61872086-61872086 on Build GRCh38
rs781412547 Chr.2: 61872075-61872075 on Build GRCh38
rs557404693 Chr.2: 61871979-61871979 on Build GRCh38
rs539300120 Chr.2: 61871957-61871957 on Build GRCh38
rs768251607 Chr.2: 61872104-61872104 on Build GRCh38

View Full Product Details