Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20159999-20160242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBX1
Entrez Gene ID
120237
Gene Name
developing brain homeobox 1
Gene Aliases
-
UniGene
Hs.558604
Gene Chromosome Location
Chr.11: 20156214-20160324 on Build GRCh38
Transcripts
NM_001029865.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604424 c.277G>T p.G93C Chr.11: 20160048-20160048 on Build GRCh38
3868971 c.310G>A p.E104K Chr.11: 20160015-20160015 on Build GRCh38
5569056 c.314C>T p.T105M Chr.11: 20160011-20160011 on Build GRCh38
1241061 c.284C>T p.P95L Chr.11: 20160041-20160041 on Build GRCh38
5714325 c.233G>A p.G78E Chr.11: 20160092-20160092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374564065 Chr.11: 20160071-20160071 on Build GRCh38
rs764491895 Chr.11: 20160040-20160040 on Build GRCh38
rs546983590 Chr.11: 20160189-20160189 on Build GRCh38
rs757076164 Chr.11: 20160144-20160144 on Build GRCh38
rs752562311 Chr.11: 20160179-20160179 on Build GRCh38
rs762018047 Chr.11: 20160214-20160214 on Build GRCh38
rs913846529 Chr.11: 20160153-20160153 on Build GRCh38
rs931757221 Chr.11: 20160129-20160129 on Build GRCh38
rs530079887 Chr.11: 20160074-20160074 on Build GRCh38
rs370098161 Chr.11: 20160056-20160056 on Build GRCh38
rs777788098 Chr.11: 20160103-20160103 on Build GRCh38
rs749139668 Chr.11: 20160018-20160018 on Build GRCh38
rs776824973 Chr.11: 20160045-20160045 on Build GRCh38
rs770437698 Chr.11: 20160020-20160020 on Build GRCh38
rs752648547 Chr.11: 20160090-20160090 on Build GRCh38
rs539147371 Chr.11: 20160223-20160223 on Build GRCh38
rs755964644 Chr.11: 20160099-20160099 on Build GRCh38
rs750679549 Chr.11: 20160063-20160063 on Build GRCh38
rs770351067 Chr.11: 20160216-20160216 on Build GRCh38
rs778529888 Chr.11: 20160164-20160164 on Build GRCh38
rs189086023 Chr.11: 20160195-20160195 on Build GRCh38
rs761139001 Chr.11: 20160039-20160039 on Build GRCh38
rs771551196 Chr.11: 20160174-20160174 on Build GRCh38
rs199851463 Chr.11: 20160059-20160059 on Build GRCh38
rs758549809 Chr.11: 20160069-20160069 on Build GRCh38
rs748974166 Chr.11: 20160118-20160118 on Build GRCh38
rs774002093 Chr.11: 20160023-20160023 on Build GRCh38
rs954904069 Chr.11: 20160110-20160110 on Build GRCh38
rs745307504 Chr.11: 20160037-20160037 on Build GRCh38
rs746563592 Chr.11: 20160180-20160180 on Build GRCh38
rs1035974405 Chr.11: 20160191-20160191 on Build GRCh38
rs745575231 Chr.11: 20160172-20160172 on Build GRCh38
rs945315972 Chr.11: 20160162-20160162 on Build GRCh38

View Full Product Details