Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92052305-92052530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB3
Entrez Gene ID
22849
Gene Name
cytoplasmic polyadenylation element binding protein 3
Gene Aliases
-
UniGene
Hs.131683
Gene Chromosome Location
Chr.10: 92046692-92291118 on Build GRCh38
Transcripts
NM_001178137.1 NM_014912.4 XM_005269630.4 XM_006717714.3 XM_006717715.3 XM_011539514.2 XM_011539515.2 XM_011539517.2 XM_011539519.2 XM_011539520.2 XM_017015927.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329733 c.1894G>T p.G632W Chr.10: 92052373-92052373 on Build GRCh38
1603853 c.1959T>C p.C653C Chr.10: 92052350-92052350 on Build GRCh38
6339707 c.1986C>A p.Y662* Chr.10: 92052323-92052323 on Build GRCh38
329271 c.1947C>A p.A649A Chr.10: 92052362-92052362 on Build GRCh38
4182208 c.1942T>C p.Y648H Chr.10: 92052325-92052325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148506777 Chr.10: 92052416-92052416 on Build GRCh38
rs568532996 Chr.10: 92052415-92052415 on Build GRCh38
rs985798921 Chr.10: 92052443-92052443 on Build GRCh38
rs371412431 Chr.10: 92052453-92052453 on Build GRCh38
rs765854491 Chr.10: 92052332-92052332 on Build GRCh38
rs759091603 Chr.10: 92052459-92052459 on Build GRCh38
rs771871745 Chr.10: 92052473-92052473 on Build GRCh38
rs759325421 Chr.10: 92052333-92052333 on Build GRCh38
rs543956067 Chr.10: 92052344-92052344 on Build GRCh38
rs767243674 Chr.10: 92052343-92052343 on Build GRCh38
rs780239545 Chr.10: 92052386-92052386 on Build GRCh38
rs537424514 Chr.10: 92052445-92052445 on Build GRCh38
rs755234133 Chr.10: 92052408-92052408 on Build GRCh38
rs777579239 Chr.10: 92052428-92052428 on Build GRCh38
rs916340155 Chr.10: 92052425-92052425 on Build GRCh38
rs758423478 Chr.10: 92052373-92052373 on Build GRCh38
rs769541702 Chr.10: 92052419-92052419 on Build GRCh38
rs762272170 Chr.10: 92052482-92052482 on Build GRCh38
rs147464061 Chr.10: 92052359-92052359 on Build GRCh38
rs557602511 Chr.10: 92052478-92052478 on Build GRCh38
rs967928970 Chr.10: 92052427-92052427 on Build GRCh38
rs750874591 Chr.10: 92052362-92052362 on Build GRCh38
rs751587481 Chr.10: 92052389-92052389 on Build GRCh38
rs976206475 Chr.10: 92052495-92052495 on Build GRCh38
rs757646182 Chr.10: 92052345-92052345 on Build GRCh38

View Full Product Details