Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 45129693-45129964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP3
Entrez Gene ID
2287
Gene Name
FK506 binding protein 3
Gene Aliases
FKBP-25, FKBP-3, FKBP25, PPIase
UniGene
Hs.509226
Gene Chromosome Location
Chr.14: 45115599-45134529 on Build GRCh38
Transcripts
NM_002013.3 XM_011536565.2 XM_017021092.1 XM_017021093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582271 c.289G>A p.E97K Chr.14: 45129823-45129823 on Build GRCh38
1477572 c.259A>C p.N87H Chr.14: 45129853-45129853 on Build GRCh38
308033 c.235A>G p.S79G Chr.14: 45129877-45129877 on Build GRCh38
6562585 c.301G>C p.E101Q Chr.14: 45129811-45129811 on Build GRCh38
5987908 c.239A>C p.K80T Chr.14: 45129873-45129873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550487416 Chr.14: 45129725-45129725 on Build GRCh38
rs777244586 Chr.14: 45129936-45129936 on Build GRCh38
rs769272889 Chr.14: 45129848-45129848 on Build GRCh38
rs1005645877 Chr.14: 45129904-45129905 on Build GRCh38
rs373710471 Chr.14: 45129743-45129743 on Build GRCh38
rs766416899 Chr.14: 45129920-45129920 on Build GRCh38
rs752941360 Chr.14: 45129931-45129931 on Build GRCh38
rs969944428 Chr.14: 45129719-45129719 on Build GRCh38
rs758477659 Chr.14: 45129749-45129749 on Build GRCh38
rs1039941424 Chr.14: 45129765-45129769 on Build GRCh38
rs748770594 Chr.14: 45129863-45129863 on Build GRCh38
rs780907496 Chr.14: 45129776-45129779 on Build GRCh38
rs569744318 Chr.14: 45129742-45129742 on Build GRCh38
rs748605260 Chr.14: 45129846-45129846 on Build GRCh38
rs981144678 Chr.14: 45129721-45129721 on Build GRCh38
rs755521526 Chr.14: 45129825-45129825 on Build GRCh38
rs376264225 Chr.14: 45129915-45129915 on Build GRCh38
rs756536964 Chr.14: 45129934-45129934 on Build GRCh38
rs369926132 Chr.14: 45129746-45129746 on Build GRCh38
rs774055821 Chr.14: 45129869-45129869 on Build GRCh38
rs139935441 Chr.14: 45129858-45129858 on Build GRCh38
rs751706871 Chr.14: 45129922-45129922 on Build GRCh38
rs780497333 Chr.14: 45129785-45129785 on Build GRCh38
rs562896633 Chr.14: 45129904-45129904 on Build GRCh38
rs767554768 Chr.14: 45129885-45129885 on Build GRCh38
rs772124958 Chr.14: 45129880-45129880 on Build GRCh38
rs908933761 Chr.14: 45129882-45129882 on Build GRCh38
rs765309859 Chr.14: 45129905-45129905 on Build GRCh38
rs62000271 Chr.14: 45129876-45129876 on Build GRCh38
rs201331434 Chr.14: 45129830-45129830 on Build GRCh38
rs747534793 Chr.14: 45129807-45129807 on Build GRCh38
rs183514536 Chr.14: 45129867-45129867 on Build GRCh38
rs772926463 Chr.14: 45129849-45129849 on Build GRCh38
rs758422509 Chr.14: 45129918-45129918 on Build GRCh38
rs928313818 Chr.14: 45129726-45129726 on Build GRCh38
rs915151168 Chr.14: 45129735-45129735 on Build GRCh38
rs750412623 Chr.14: 45129748-45129748 on Build GRCh38
rs755428499 Chr.14: 45129923-45129923 on Build GRCh38
rs201108052 Chr.14: 45129836-45129836 on Build GRCh38
rs748184229 Chr.14: 45129864-45129864 on Build GRCh38
rs781729063 Chr.14: 45129930-45129930 on Build GRCh38
rs775130834 Chr.14: 45129900-45129900 on Build GRCh38
rs760527701 Chr.14: 45129901-45129901 on Build GRCh38
rs766488256 Chr.14: 45129918-45129919 on Build GRCh38
rs781407019 Chr.14: 45129832-45129832 on Build GRCh38

View Full Product Details