Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 26566945-26567214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO3
Entrez Gene ID
63982
Gene Name
anoctamin 3
Gene Aliases
C11orf25, DYT23, DYT24, GENX-3947, TMEM16C
UniGene
Hs.577269
Gene Chromosome Location
Chr.11: 26189123-26663289 on Build GRCh38
Transcripts
NM_001313726.1 NM_001313727.1 NM_031418.3 XM_017018118.1 XM_017018119.1 NM_001135091.1 NM_001135092.1 NM_145650.3 XM_017017235.1 XM_017017236.1

Gene Symbol
MUC15
Entrez Gene ID
143662
Gene Name
mucin 15, cell surface associated
Gene Aliases
MUC-15, PAS3, PASIII
UniGene
Hs.407152
Gene Chromosome Location
Chr.11: 26559032-26572268 on Build GRCh38
Transcripts
NM_001313726.1 NM_001313727.1 NM_031418.3 XM_017018118.1 XM_017018119.1 NM_001135091.1 NM_001135092.1 NM_145650.3 XM_017017235.1 XM_017017236.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5783143 c.43+9C>A p.? Chr.11: 26567043-26567043 on Build GRCh38
1728511 c.4G>A p.G2S Chr.11: 26567091-26567091 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs910782425 Chr.11: 26567058-26567058 on Build GRCh38
rs865885555 Chr.11: 26567090-26567090 on Build GRCh38
rs933288024 Chr.11: 26566986-26566986 on Build GRCh38
rs929110575 Chr.11: 26567168-26567168 on Build GRCh38
rs147213313 Chr.11: 26567044-26567044 on Build GRCh38
rs377215628 Chr.11: 26567055-26567055 on Build GRCh38
rs569918362 Chr.11: 26567129-26567129 on Build GRCh38
rs986316682 Chr.11: 26567036-26567036 on Build GRCh38
rs930096233 Chr.11: 26567171-26567171 on Build GRCh38
rs1037903974 Chr.11: 26567082-26567082 on Build GRCh38
rs922863320 Chr.11: 26567153-26567153 on Build GRCh38
rs967819592 Chr.11: 26567027-26567027 on Build GRCh38
rs566671441 Chr.11: 26567079-26567079 on Build GRCh38
rs750807191 Chr.11: 26567078-26567078 on Build GRCh38
rs753301751 Chr.11: 26567008-26567008 on Build GRCh38
rs897981488 Chr.11: 26567097-26567097 on Build GRCh38
rs368209928 Chr.11: 26567076-26567076 on Build GRCh38

View Full Product Details