Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37183953-37184226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPN2
Entrez Gene ID
6185
Gene Name
ribophorin II
Gene Aliases
RIBIIR, RPN-II, RPNII, SWP1
UniGene
Hs.370895
Gene Chromosome Location
Chr.20: 37179046-37241623 on Build GRCh38
Transcripts
NM_001135771.2 NM_001324299.1 NM_001324301.1 NM_001324302.1 NM_001324303.1 NM_001324304.1 NM_001324305.1 NM_001324306.1 NM_002951.4 XM_006723851.2 XM_006723852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4394058 c.34T>C p.L12L Chr.20: 37184200-37184200 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760195435 Chr.20: 37184184-37184184 on Build GRCh38
rs571010809 Chr.20: 37184069-37184069 on Build GRCh38
rs770485866 Chr.20: 37184174-37184174 on Build GRCh38
rs969687030 Chr.20: 37184061-37184061 on Build GRCh38
rs775238433 Chr.20: 37184176-37184176 on Build GRCh38
rs757120224 Chr.20: 37184134-37184134 on Build GRCh38
rs530180337 Chr.20: 37184041-37184041 on Build GRCh38
rs969026887 Chr.20: 37184082-37184082 on Build GRCh38
rs547959692 Chr.20: 37184055-37184055 on Build GRCh38
rs201610870 Chr.20: 37184171-37184171 on Build GRCh38
rs750576659 Chr.20: 37184162-37184162 on Build GRCh38
rs747161354 Chr.20: 37184167-37184167 on Build GRCh38
rs1037594748 Chr.20: 37184122-37184122 on Build GRCh38
rs1027049961 Chr.20: 37184056-37184056 on Build GRCh38
rs748666082 Chr.20: 37184173-37184173 on Build GRCh38
rs77007680 Chr.20: 37184071-37184071 on Build GRCh38
rs912644765 Chr.20: 37184106-37184106 on Build GRCh38
rs375317516 Chr.20: 37184203-37184203 on Build GRCh38
rs552076360 Chr.20: 37184120-37184120 on Build GRCh38
rs375013035 Chr.20: 37184148-37184148 on Build GRCh38
rs774981812 Chr.20: 37184119-37184119 on Build GRCh38
rs753794559 Chr.20: 37184133-37184133 on Build GRCh38
rs932620970 Chr.20: 37184096-37184096 on Build GRCh38
rs995214124 Chr.20: 37184044-37184044 on Build GRCh38
rs766774507 Chr.20: 37184177-37184177 on Build GRCh38
rs772220569 Chr.20: 37184110-37184110 on Build GRCh38
rs1049771916 Chr.20: 37183987-37183987 on Build GRCh38
rs765483448 Chr.20: 37184084-37184084 on Build GRCh38
rs16986470 Chr.20: 37184123-37184123 on Build GRCh38
rs758387019 Chr.20: 37184163-37184163 on Build GRCh38
rs1008774326 Chr.20: 37184007-37184007 on Build GRCh38
rs536913734 Chr.20: 37184079-37184079 on Build GRCh38
rs112682509 Chr.20: 37184178-37184178 on Build GRCh38
rs745726528 Chr.20: 37184015-37184015 on Build GRCh38
rs925300714 Chr.20: 37184089-37184089 on Build GRCh38
rs780366243 Chr.20: 37184164-37184164 on Build GRCh38
rs989048216 Chr.20: 37184072-37184072 on Build GRCh38
rs757255073 Chr.20: 37184129-37184130 on Build GRCh38
rs760807826 Chr.20: 37184131-37184131 on Build GRCh38
rs1001295561 Chr.20: 37184062-37184062 on Build GRCh38
rs891301632 Chr.20: 37184001-37184001 on Build GRCh38
rs985514058 Chr.20: 37184097-37184097 on Build GRCh38
rs199630624 Chr.20: 37184175-37184175 on Build GRCh38
rs1037327329 Chr.20: 37184027-37184027 on Build GRCh38
rs763881413 Chr.20: 37184190-37184190 on Build GRCh38
rs767254529 Chr.20: 37184117-37184117 on Build GRCh38
rs754101577 Chr.20: 37184199-37184199 on Build GRCh38

View Full Product Details