Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19743011-19743282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH3A1
Entrez Gene ID
218
Gene Name
aldehyde dehydrogenase 3 family member A1
Gene Aliases
ALDH3, ALDHIII
UniGene
Hs.531682
Gene Chromosome Location
Chr.17: 19737984-19748433 on Build GRCh38
Transcripts
NM_000691.4 NM_001135167.1 NM_001135168.1 XM_005256522.1 XM_005256523.2 XM_005256524.3 XM_011523730.1 XM_011523731.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
976452 c.389C>T p.A130V Chr.17: 19743237-19743237 on Build GRCh38
2798331 c.394G>T p.G132W Chr.17: 19743232-19743232 on Build GRCh38
3515076 c.356A>G p.N119S Chr.17: 19743270-19743270 on Build GRCh38
3515075 c.392C>T p.A131V Chr.17: 19743234-19743234 on Build GRCh38
5624351 c.388G>C p.A130P Chr.17: 19743238-19743238 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950473030 Chr.17: 19743069-19743069 on Build GRCh38
rs34474118 Chr.17: 19743186-19743187 on Build GRCh38
rs1008555761 Chr.17: 19743141-19743141 on Build GRCh38
rs775753026 Chr.17: 19743249-19743249 on Build GRCh38
rs781475117 Chr.17: 19743186-19743186 on Build GRCh38
rs376465259 Chr.17: 19743183-19743183 on Build GRCh38
rs759203617 Chr.17: 19743210-19743210 on Build GRCh38
rs370374404 Chr.17: 19743197-19743197 on Build GRCh38
rs536106180 Chr.17: 19743191-19743191 on Build GRCh38
rs749526257 Chr.17: 19743243-19743243 on Build GRCh38
rs757352910 Chr.17: 19743179-19743179 on Build GRCh38
rs751544540 Chr.17: 19743156-19743156 on Build GRCh38
rs924684896 Chr.17: 19743215-19743215 on Build GRCh38
rs150970681 Chr.17: 19743239-19743239 on Build GRCh38
rs995051882 Chr.17: 19743042-19743042 on Build GRCh38
rs200199114 Chr.17: 19743245-19743245 on Build GRCh38
rs771232956 Chr.17: 19743044-19743044 on Build GRCh38
rs1044535360 Chr.17: 19743031-19743031 on Build GRCh38
rs201715060 Chr.17: 19743188-19743188 on Build GRCh38
rs201166581 Chr.17: 19743194-19743194 on Build GRCh38
rs759178925 Chr.17: 19743130-19743130 on Build GRCh38
rs1056981728 Chr.17: 19743057-19743057 on Build GRCh38
rs757328458 Chr.17: 19743231-19743231 on Build GRCh38
rs751526409 Chr.17: 19743227-19743227 on Build GRCh38
rs938535179 Chr.17: 19743055-19743055 on Build GRCh38
rs756253107 Chr.17: 19743238-19743238 on Build GRCh38
rs548157710 Chr.17: 19743226-19743226 on Build GRCh38
rs117328009 Chr.17: 19743208-19743208 on Build GRCh38
rs777811199 Chr.17: 19743145-19743145 on Build GRCh38
rs762768333 Chr.17: 19743225-19743225 on Build GRCh38
rs779319405 Chr.17: 19743253-19743253 on Build GRCh38
rs536804477 Chr.17: 19743244-19743244 on Build GRCh38
rs1008784133 Chr.17: 19743262-19743262 on Build GRCh38
rs764747434 Chr.17: 19743212-19743212 on Build GRCh38
rs758287966 Chr.17: 19743142-19743142 on Build GRCh38
rs770423248 Chr.17: 19743198-19743198 on Build GRCh38
rs376130132 Chr.17: 19743232-19743232 on Build GRCh38
rs145896313 Chr.17: 19743193-19743193 on Build GRCh38
rs915193266 Chr.17: 19743202-19743202 on Build GRCh38
rs750423189 Chr.17: 19743236-19743236 on Build GRCh38
rs769016246 Chr.17: 19743195-19743195 on Build GRCh38
rs765049018 Chr.17: 19743137-19743137 on Build GRCh38
rs752483408 Chr.17: 19743138-19743138 on Build GRCh38
rs899296012 Chr.17: 19743030-19743030 on Build GRCh38

View Full Product Details