Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 46679896-46680169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST8SIA5
Entrez Gene ID
29906
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5
Gene Aliases
SIAT8-E, SIAT8E, ST8SiaV
UniGene
Hs.465025
Gene Chromosome Location
Chr.18: 46679118-46757169 on Build GRCh38
Transcripts
NM_001307986.1 NM_001307987.1 NM_013305.5 XM_017025715.1 XM_017025716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
98152 c.1088G>A p.R363Q Chr.18: 46680085-46680085 on Build GRCh38
6333437 c.1099C>A p.R367S Chr.18: 46680074-46680074 on Build GRCh38
4072314 c.1046C>G p.A349G Chr.18: 46680127-46680127 on Build GRCh38
1266871 c.1025A>C p.K342T Chr.18: 46680148-46680148 on Build GRCh38
5388170 c.1051C>T p.P351S Chr.18: 46680122-46680122 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529675198 Chr.18: 46680020-46680020 on Build GRCh38
rs757280313 Chr.18: 46680022-46680033 on Build GRCh38
rs370702424 Chr.18: 46680060-46680060 on Build GRCh38
rs1048804426 Chr.18: 46679992-46679992 on Build GRCh38
rs538781555 Chr.18: 46680142-46680142 on Build GRCh38
rs770230911 Chr.18: 46680014-46680014 on Build GRCh38
rs773255154 Chr.18: 46680005-46680005 on Build GRCh38
rs780097367 Chr.18: 46680064-46680064 on Build GRCh38
rs772368035 Chr.18: 46679999-46679999 on Build GRCh38
rs767478422 Chr.18: 46680059-46680059 on Build GRCh38
rs895534383 Chr.18: 46680112-46680112 on Build GRCh38
rs752844475 Chr.18: 46680078-46680078 on Build GRCh38
rs376358845 Chr.18: 46680106-46680106 on Build GRCh38
rs775695315 Chr.18: 46680042-46680042 on Build GRCh38
rs761496260 Chr.18: 46680058-46680058 on Build GRCh38
rs768093093 Chr.18: 46680127-46680127 on Build GRCh38
rs886887576 Chr.18: 46679953-46679953 on Build GRCh38
rs758454262 Chr.18: 46680079-46680079 on Build GRCh38
rs1001225199 Chr.18: 46680071-46680071 on Build GRCh38
rs776980061 Chr.18: 46680009-46680009 on Build GRCh38
rs1022758106 Chr.18: 46680097-46680097 on Build GRCh38
rs777875690 Chr.18: 46680081-46680081 on Build GRCh38
rs536769837 Chr.18: 46680041-46680041 on Build GRCh38
rs781553691 Chr.18: 46680104-46680104 on Build GRCh38
rs761384435 Chr.18: 46680139-46680139 on Build GRCh38
rs759748191 Chr.18: 46679965-46679965 on Build GRCh38
rs1017028045 Chr.18: 46680011-46680011 on Build GRCh38
rs774988557 Chr.18: 46680016-46680016 on Build GRCh38
rs896984196 Chr.18: 46679945-46679945 on Build GRCh38
rs773908092 Chr.18: 46680136-46680136 on Build GRCh38
rs763496347 Chr.18: 46680117-46680117 on Build GRCh38
rs765551790 Chr.18: 46680010-46680010 on Build GRCh38
rs751429405 Chr.18: 46680021-46680033 on Build GRCh38
rs746167460 Chr.18: 46680105-46680105 on Build GRCh38
rs1040823386 Chr.18: 46679927-46679927 on Build GRCh38
rs750272677 Chr.18: 46680146-46680146 on Build GRCh38
rs756123767 Chr.18: 46680063-46680063 on Build GRCh38
rs751220976 Chr.18: 46680057-46680057 on Build GRCh38
rs373700133 Chr.18: 46680085-46680085 on Build GRCh38
rs747257419 Chr.18: 46680083-46680083 on Build GRCh38
rs771176027 Chr.18: 46680102-46680102 on Build GRCh38
rs373039246 Chr.18: 46679947-46679947 on Build GRCh38
rs924684866 Chr.18: 46680138-46680138 on Build GRCh38
rs147438446 Chr.18: 46680114-46680114 on Build GRCh38
rs1003862599 Chr.18: 46680001-46680001 on Build GRCh38
rs560523484 Chr.18: 46679922-46679922 on Build GRCh38
rs201780051 Chr.18: 46680027-46680027 on Build GRCh38
rs971201898 Chr.18: 46680098-46680098 on Build GRCh38

View Full Product Details