Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122929916-122930171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
UniGene
Hs.511951
Gene Chromosome Location
Chr.12: 122918708-122975160 on Build GRCh38
Transcripts
NM_001243013.1 NM_001243014.1 NM_019624.3 NM_019625.3 NM_203444.3 XM_005253558.2 XM_011538095.2 XM_011538096.2 XM_011538098.2 XM_017019103.1 XM_017019104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1359703 c.2086A>G p.I696V Chr.12: 122930126-122930126 on Build GRCh38
5374477 c.2043C>T p.I681I Chr.12: 122930169-122930169 on Build GRCh38
5374476 c.2173C>T p.Q725* Chr.12: 122930039-122930039 on Build GRCh38
1359702 c.2167A>G p.T723A Chr.12: 122930045-122930045 on Build GRCh38
4988515 c.2150G>A p.R717H Chr.12: 122930062-122930062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148914003 Chr.12: 122930057-122930057 on Build GRCh38
rs138175647 Chr.12: 122930069-122930069 on Build GRCh38
rs141512966 Chr.12: 122930128-122930128 on Build GRCh38
rs756487180 Chr.12: 122930133-122930133 on Build GRCh38
rs777887261 Chr.12: 122929953-122929953 on Build GRCh38
rs897039608 Chr.12: 122930058-122930058 on Build GRCh38
rs770338952 Chr.12: 122929971-122929971 on Build GRCh38
rs566484773 Chr.12: 122929968-122929968 on Build GRCh38
rs1038273184 Chr.12: 122930046-122930046 on Build GRCh38
rs777845065 Chr.12: 122930035-122930035 on Build GRCh38
rs548262950 Chr.12: 122929965-122929965 on Build GRCh38
rs750598545 Chr.12: 122930121-122930121 on Build GRCh38
rs765074033 Chr.12: 122930094-122930094 on Build GRCh38
rs765938649 Chr.12: 122930000-122930000 on Build GRCh38
rs369954984 Chr.12: 122929967-122929967 on Build GRCh38
rs145637805 Chr.12: 122930060-122930060 on Build GRCh38
rs556899913 Chr.12: 122930103-122930103 on Build GRCh38
rs763016891 Chr.12: 122930104-122930104 on Build GRCh38
rs757558895 Chr.12: 122930052-122930052 on Build GRCh38
rs911050418 Chr.12: 122930022-122930022 on Build GRCh38
rs939043612 Chr.12: 122929966-122929966 on Build GRCh38
rs763925609 Chr.12: 122930112-122930112 on Build GRCh38
rs915153837 Chr.12: 122930095-122930095 on Build GRCh38
rs751587429 Chr.12: 122930040-122930040 on Build GRCh38
rs1045335406 Chr.12: 122929944-122929944 on Build GRCh38
rs370679674 Chr.12: 122930091-122930091 on Build GRCh38
rs1047343324 Chr.12: 122930019-122930019 on Build GRCh38
rs1023837402 Chr.12: 122930072-122930072 on Build GRCh38
rs770106220 Chr.12: 122930061-122930061 on Build GRCh38
rs781428831 Chr.12: 122930118-122930118 on Build GRCh38
rs189653462 Chr.12: 122929994-122929994 on Build GRCh38
rs772750441 Chr.12: 122929993-122929993 on Build GRCh38
rs997728616 Chr.12: 122930102-122930102 on Build GRCh38
rs758471224 Chr.12: 122929952-122929952 on Build GRCh38
rs538504962 Chr.12: 122930092-122930092 on Build GRCh38
rs993187105 Chr.12: 122930008-122930008 on Build GRCh38
rs775949751 Chr.12: 122929980-122929980 on Build GRCh38
rs771564666 Chr.12: 122929987-122929987 on Build GRCh38
rs148331618 Chr.12: 122930120-122930120 on Build GRCh38
rs749377058 Chr.12: 122929939-122929948 on Build GRCh38
rs752640304 Chr.12: 122930018-122930018 on Build GRCh38
rs373159982 Chr.12: 122929970-122929970 on Build GRCh38
rs772841474 Chr.12: 122930076-122930076 on Build GRCh38
rs976027877 Chr.12: 122929979-122929979 on Build GRCh38
rs778980983 Chr.12: 122929947-122929947 on Build GRCh38
rs747197529 Chr.12: 122929958-122929958 on Build GRCh38
rs765304579 Chr.12: 122930014-122930014 on Build GRCh38
rs751711542 Chr.12: 122930114-122930114 on Build GRCh38
rs749742353 Chr.12: 122930063-122930063 on Build GRCh38
rs529640877 Chr.12: 122929934-122929934 on Build GRCh38
rs759382689 Chr.12: 122930010-122930010 on Build GRCh38
rs761346615 Chr.12: 122929986-122929986 on Build GRCh38
rs770665149 Chr.12: 122930085-122930085 on Build GRCh38
rs748287773 Chr.12: 122929950-122929950 on Build GRCh38
rs142777722 Chr.12: 122930078-122930078 on Build GRCh38
rs968335675 Chr.12: 122930113-122930113 on Build GRCh38
rs145038250 Chr.12: 122930116-122930116 on Build GRCh38
rs375922544 Chr.12: 122929943-122929943 on Build GRCh38
rs376322922 Chr.12: 122930062-122930062 on Build GRCh38
rs758423439 Chr.12: 122930031-122930031 on Build GRCh38
rs372144621 Chr.12: 122930009-122930009 on Build GRCh38
rs1001463067 Chr.12: 122930090-122930090 on Build GRCh38
rs148691993 Chr.12: 122930148-122930148 on Build GRCh38

View Full Product Details