Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 245081892-245082165 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB2
Entrez Gene ID
84288
Gene Name
EF-hand calcium binding domain 2
Gene Aliases
CFAP200, DRC8
UniGene
Hs.134857
Gene Chromosome Location
Chr.1: 244969739-245125228 on Build GRCh38
Transcripts
NM_001143943.1 NM_001290327.1 NM_032328.3 NR_026587.1 XM_011544300.2 XM_011544301.1 XM_011544302.2 XM_017002538.1 XM_017002539.1 XM_017002540.1 XM_017002541.1 XM_017002542.1 XM_017002543.1 XM_017002544.1 XM_017002545.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5380316 c.637C>T p.L213F Chr.1: 245082128-245082128 on Build GRCh38
7365878 c.233C>T p.P78L Chr.1: 245082132-245082132 on Build GRCh38
2051556 c.220G>A p.E74K Chr.1: 245082119-245082119 on Build GRCh38
7136218 c.215G>T p.R72L Chr.1: 245082114-245082114 on Build GRCh38
1748277 c.256G>C p.E86Q Chr.1: 245082155-245082155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966401988 Chr.1: 245082010-245082010 on Build GRCh38
rs773087834 Chr.1: 245082114-245082114 on Build GRCh38
rs779387683 Chr.1: 245082038-245082038 on Build GRCh38
rs767268130 Chr.1: 245082133-245082133 on Build GRCh38
rs761571533 Chr.1: 245082106-245082106 on Build GRCh38
rs750388985 Chr.1: 245082131-245082131 on Build GRCh38
rs544391259 Chr.1: 245081968-245081968 on Build GRCh38
rs557402578 Chr.1: 245082119-245082119 on Build GRCh38
rs974542518 Chr.1: 245082077-245082077 on Build GRCh38
rs780056053 Chr.1: 245082049-245082049 on Build GRCh38
rs529930682 Chr.1: 245082054-245082054 on Build GRCh38
rs142888002 Chr.1: 245082118-245082118 on Build GRCh38
rs749801218 Chr.1: 245082072-245082072 on Build GRCh38
rs561075630 Chr.1: 245082021-245082021 on Build GRCh38
rs757892084 Chr.1: 245082032-245082032 on Build GRCh38
rs778324389 Chr.1: 245082071-245082071 on Build GRCh38
rs764978665 Chr.1: 245082113-245082113 on Build GRCh38
rs1054899950 Chr.1: 245081939-245081939 on Build GRCh38
rs1002289011 Chr.1: 245081919-245081919 on Build GRCh38
rs1008825530 Chr.1: 245082033-245082033 on Build GRCh38
rs893519997 Chr.1: 245081967-245081967 on Build GRCh38
rs751417929 Chr.1: 245082089-245082091 on Build GRCh38
rs759617162 Chr.1: 245082084-245082084 on Build GRCh38
rs984218808 Chr.1: 245082109-245082109 on Build GRCh38
rs7551261 Chr.1: 245082132-245082132 on Build GRCh38
rs908570524 Chr.1: 245082111-245082111 on Build GRCh38
rs751133616 Chr.1: 245082051-245082051 on Build GRCh38
rs769745280 Chr.1: 245082128-245082128 on Build GRCh38
rs751582223 Chr.1: 245081926-245081926 on Build GRCh38
rs375113841 Chr.1: 245082092-245082092 on Build GRCh38
rs748522930 Chr.1: 245082062-245082062 on Build GRCh38
rs10927387 Chr.1: 245082100-245082100 on Build GRCh38
rs575612193 Chr.1: 245081945-245081945 on Build GRCh38
rs1021065924 Chr.1: 245081981-245081981 on Build GRCh38
rs10927386 Chr.1: 245082076-245082076 on Build GRCh38

View Full Product Details