Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27329144-27329381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C2
Entrez Gene ID
2976
Gene Name
general transcription factor IIIC subunit 2
Gene Aliases
TFIIIC-BETA, TFIIIC110
UniGene
Hs.745168
Gene Chromosome Location
Chr.2: 27325849-27357034 on Build GRCh38
Transcripts
NM_001035521.2 NM_001318909.1 NM_001521.3 XM_005264272.4 XM_011532801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4405909 c.1969T>G p.L657V Chr.2: 27329191-27329191 on Build GRCh38
6334120 c.1941C>A p.Y647* Chr.2: 27329219-27329219 on Build GRCh38
4164495 c.1951A>T p.N651Y Chr.2: 27329209-27329209 on Build GRCh38
6676818 c.1930C>T p.R644* Chr.2: 27329230-27329230 on Build GRCh38
4689191 c.1917A>C p.K639N Chr.2: 27329243-27329243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747775019 Chr.2: 27329227-27329227 on Build GRCh38
rs896537055 Chr.2: 27329207-27329207 on Build GRCh38
rs778809914 Chr.2: 27329342-27329342 on Build GRCh38
rs776853502 Chr.2: 27329299-27329299 on Build GRCh38
rs752023228 Chr.2: 27329286-27329287 on Build GRCh38
rs572927438 Chr.2: 27329349-27329349 on Build GRCh38
rs187061145 Chr.2: 27329322-27329322 on Build GRCh38
rs763423156 Chr.2: 27329315-27329315 on Build GRCh38
rs765304322 Chr.2: 27329199-27329199 on Build GRCh38
rs750470401 Chr.2: 27329313-27329313 on Build GRCh38
rs533881871 Chr.2: 27329356-27329356 on Build GRCh38
rs752023144 Chr.2: 27329317-27329317 on Build GRCh38
rs762029935 Chr.2: 27329305-27329305 on Build GRCh38
rs143400550 Chr.2: 27329204-27329204 on Build GRCh38
rs750901561 Chr.2: 27329203-27329203 on Build GRCh38
rs749203416 Chr.2: 27329276-27329276 on Build GRCh38
rs568136490 Chr.2: 27329246-27329246 on Build GRCh38
rs201499584 Chr.2: 27329278-27329278 on Build GRCh38
rs550971239 Chr.2: 27329294-27329294 on Build GRCh38
rs964522837 Chr.2: 27329347-27329347 on Build GRCh38
rs748799655 Chr.2: 27329327-27329327 on Build GRCh38
rs774171667 Chr.2: 27329287-27329287 on Build GRCh38
rs138720572 Chr.2: 27329219-27329219 on Build GRCh38
rs367917139 Chr.2: 27329288-27329288 on Build GRCh38
rs775317006 Chr.2: 27329355-27329355 on Build GRCh38
rs777296231 Chr.2: 27329258-27329258 on Build GRCh38
rs780974234 Chr.2: 27329226-27329226 on Build GRCh38
rs183210129 Chr.2: 27329196-27329196 on Build GRCh38
rs569568286 Chr.2: 27329321-27329321 on Build GRCh38
rs558376671 Chr.2: 27329323-27329323 on Build GRCh38
rs769189061 Chr.2: 27329234-27329234 on Build GRCh38
rs757252797 Chr.2: 27329329-27329329 on Build GRCh38
rs751953553 Chr.2: 27329218-27329218 on Build GRCh38
rs777195510 Chr.2: 27329324-27329324 on Build GRCh38

View Full Product Details