Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36088693-36088928 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL3
Entrez Gene ID
6348
Gene Name
C-C motif chemokine ligand 3
Gene Aliases
G0S19-1, LD78ALPHA, MIP-1-alpha, MIP1A, SCYA3
UniGene
Hs.514107
Gene Chromosome Location
Chr.17: 36088256-36090160 on Build GRCh38
Transcripts
NM_002983.2

SNP Detail

SNP ID Chromosome Location
rs34171309 Chr.17: 36088717-36088717 on Build GRCh38
rs555235636 Chr.17: 36088809-36088809 on Build GRCh38
rs764051613 Chr.17: 36088771-36088771 on Build GRCh38
rs779590579 Chr.17: 36088737-36088737 on Build GRCh38
rs753553385 Chr.17: 36088791-36088791 on Build GRCh38
rs759898496 Chr.17: 36088814-36088814 on Build GRCh38
rs578119387 Chr.17: 36088873-36088873 on Build GRCh38
rs755625112 Chr.17: 36088781-36088781 on Build GRCh38
rs540710731 Chr.17: 36088833-36088833 on Build GRCh38
rs369914435 Chr.17: 36088777-36088777 on Build GRCh38
rs147323689 Chr.17: 36088733-36088733 on Build GRCh38
rs779583503 Chr.17: 36088790-36088790 on Build GRCh38
rs1719130 Chr.17: 36088900-36088900 on Build GRCh38
rs897655273 Chr.17: 36088824-36088824 on Build GRCh38
rs5029408 Chr.17: 36088746-36088746 on Build GRCh38
rs569801296 Chr.17: 36088792-36088792 on Build GRCh38
rs998687647 Chr.17: 36088895-36088895 on Build GRCh38
rs776848484 Chr.17: 36088811-36088811 on Build GRCh38
rs748710179 Chr.17: 36088744-36088744 on Build GRCh38
rs5029409 Chr.17: 36088749-36088749 on Build GRCh38
rs1018732965 Chr.17: 36088869-36088869 on Build GRCh38
rs5029410 Chr.17: 36088805-36088805 on Build GRCh38
rs542527118 Chr.17: 36088877-36088877 on Build GRCh38
rs778166572 Chr.17: 36088803-36088803 on Build GRCh38
rs770996135 Chr.17: 36088754-36088754 on Build GRCh38
rs572302960 Chr.17: 36088832-36088832 on Build GRCh38
rs556756996 Chr.17: 36088821-36088821 on Build GRCh38
rs554356555 Chr.17: 36088834-36088834 on Build GRCh38
rs545841233 Chr.17: 36088865-36088865 on Build GRCh38
rs772441681 Chr.17: 36088800-36088800 on Build GRCh38
rs776769566 Chr.17: 36088756-36088756 on Build GRCh38
rs182552517 Chr.17: 36088886-36088886 on Build GRCh38
rs748516138 Chr.17: 36088794-36088794 on Build GRCh38
rs562905594 Chr.17: 36088870-36088870 on Build GRCh38
rs748914581 Chr.17: 36088742-36088742 on Build GRCh38
rs557795937 Chr.17: 36088883-36088883 on Build GRCh38
rs3210157 Chr.17: 36088743-36088743 on Build GRCh38
rs773703371 Chr.17: 36088748-36088748 on Build GRCh38
rs751391118 Chr.17: 36088773-36088773 on Build GRCh38
rs377681850 Chr.17: 36088761-36088761 on Build GRCh38
rs967550713 Chr.17: 36088901-36088901 on Build GRCh38
rs556132749 Chr.17: 36088780-36088780 on Build GRCh38
rs577304178 Chr.17: 36088838-36088838 on Build GRCh38

View Full Product Details