Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37211868-37212093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEBPZ
Entrez Gene ID
10153
Gene Name
CCAAT/enhancer binding protein zeta
Gene Aliases
CBF, CBF2, HSP-CBF, NOC1
UniGene
Hs.135406
Gene Chromosome Location
Chr.2: 37201629-37231597 on Build GRCh38
Transcripts
NM_005760.2 NR_136316.1 XM_017003108.1 XM_017003109.1

Gene Symbol
CEBPZOS
Entrez Gene ID
100505876
Gene Name
CEBPZ opposite strand
Gene Aliases
CEBPZ-AS1
UniGene
Hs.666958
Gene Chromosome Location
Chr.2: 37196451-37216193 on Build GRCh38
Transcripts
NM_005760.2 NR_136316.1 XM_017003108.1 XM_017003109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094094 c.2683A>G p.N895D Chr.2: 37211960-37211960 on Build GRCh38
4671348 c.2639A>G p.N880S Chr.2: 37212004-37212004 on Build GRCh38
4133828 c.2671G>T p.D891Y Chr.2: 37211972-37211972 on Build GRCh38
7184314 c.2727A>C p.E909D Chr.2: 37211916-37211916 on Build GRCh38
7193292 c.2738A>C p.E913A Chr.2: 37211905-37211905 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752604045 Chr.2: 37212052-37212052 on Build GRCh38
rs112454592 Chr.2: 37211899-37211899 on Build GRCh38
rs890365172 Chr.2: 37212046-37212046 on Build GRCh38
rs370973266 Chr.2: 37211971-37211971 on Build GRCh38
rs746008872 Chr.2: 37211933-37211933 on Build GRCh38
rs188301757 Chr.2: 37211974-37211974 on Build GRCh38
rs903248458 Chr.2: 37211953-37211953 on Build GRCh38
rs998828941 Chr.2: 37211966-37211966 on Build GRCh38
rs941542713 Chr.2: 37211986-37211986 on Build GRCh38
rs776637530 Chr.2: 37211943-37211943 on Build GRCh38
rs899921987 Chr.2: 37211997-37211997 on Build GRCh38
rs758525915 Chr.2: 37211916-37211916 on Build GRCh38
rs375786270 Chr.2: 37211949-37211949 on Build GRCh38
rs530467733 Chr.2: 37211940-37211940 on Build GRCh38
rs780294911 Chr.2: 37211932-37211932 on Build GRCh38
rs772013583 Chr.2: 37211934-37211934 on Build GRCh38
rs957290644 Chr.2: 37211995-37211995 on Build GRCh38
rs571907843 Chr.2: 37212002-37212002 on Build GRCh38
rs768578319 Chr.2: 37211942-37211942 on Build GRCh38
rs757828516 Chr.2: 37211910-37211910 on Build GRCh38
rs887306266 Chr.2: 37211935-37211935 on Build GRCh38
rs753100745 Chr.2: 37211915-37211915 on Build GRCh38
rs765393705 Chr.2: 37212053-37212053 on Build GRCh38
rs768774611 Chr.2: 37212036-37212036 on Build GRCh38
rs574810548 Chr.2: 37212035-37212035 on Build GRCh38
rs765754873 Chr.2: 37211963-37211963 on Build GRCh38
rs955257906 Chr.2: 37212042-37212042 on Build GRCh38
rs748000009 Chr.2: 37212044-37212044 on Build GRCh38
rs776690290 Chr.2: 37212041-37212041 on Build GRCh38
rs569299504 Chr.2: 37211913-37211913 on Build GRCh38
rs761491408 Chr.2: 37211894-37211894 on Build GRCh38
rs753893788 Chr.2: 37212004-37212005 on Build GRCh38
rs185440523 Chr.2: 37211958-37211958 on Build GRCh38
rs750009698 Chr.2: 37211907-37211907 on Build GRCh38
rs192503385 Chr.2: 37211948-37211948 on Build GRCh38
rs779235292 Chr.2: 37212007-37212007 on Build GRCh38
rs764979950 Chr.2: 37211902-37211902 on Build GRCh38
rs758518352 Chr.2: 37212024-37212024 on Build GRCh38
rs746036968 Chr.2: 37212021-37212021 on Build GRCh38
rs761901247 Chr.2: 37212044-37212044 on Build GRCh38
rs779933992 Chr.2: 37212032-37212032 on Build GRCh38
rs753256697 Chr.2: 37211965-37211965 on Build GRCh38
rs769440108 Chr.2: 37212062-37212062 on Build GRCh38
rs138555553 Chr.2: 37211929-37211929 on Build GRCh38
rs757594264 Chr.2: 37211993-37211993 on Build GRCh38
rs372501847 Chr.2: 37211921-37211921 on Build GRCh38
rs754257607 Chr.2: 37211988-37211988 on Build GRCh38
rs532915869 Chr.2: 37211926-37211926 on Build GRCh38
rs757541050 Chr.2: 37211927-37211927 on Build GRCh38

View Full Product Details