Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4359131-4359380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO7
Entrez Gene ID
79585
Gene Name
coronin 7
Gene Aliases
0610011B16Rik, CRN7, POD1
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4354542-4416961 on Build GRCh38
Transcripts
NM_001201472.1 NM_001201473.1 NM_024535.4 NM_001201479.1

Gene Symbol
CORO7-PAM16
Entrez Gene ID
100529144
Gene Name
CORO7-PAM16 readthrough
Gene Aliases
-
UniGene
Hs.437957
Gene Chromosome Location
Chr.16: 4340251-4416961 on Build GRCh38
Transcripts
NM_001201472.1 NM_001201473.1 NM_024535.4 NM_001201479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5484027 c.2340+1G>A p.? Chr.16: 4359295-4359295 on Build GRCh38
1580737 c.2287G>A p.E763K Chr.16: 4359349-4359349 on Build GRCh38
6197066 c.2260C>T p.R754C Chr.16: 4359376-4359376 on Build GRCh38
3402326 c.2261G>A p.R754H Chr.16: 4359375-4359375 on Build GRCh38
3509619 c.2301C>T p.F767F Chr.16: 4359335-4359335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770938661 Chr.16: 4359302-4359302 on Build GRCh38
rs936421645 Chr.16: 4359203-4359203 on Build GRCh38
rs140974533 Chr.16: 4359350-4359350 on Build GRCh38
rs1039100516 Chr.16: 4359283-4359283 on Build GRCh38
rs962936852 Chr.16: 4359220-4359220 on Build GRCh38
rs1013728383 Chr.16: 4359242-4359242 on Build GRCh38
rs1045839238 Chr.16: 4359267-4359267 on Build GRCh38
rs991866004 Chr.16: 4359232-4359232 on Build GRCh38
rs758741547 Chr.16: 4359269-4359269 on Build GRCh38
rs367550652 Chr.16: 4359279-4359279 on Build GRCh38
rs748762685 Chr.16: 4359280-4359280 on Build GRCh38
rs769625549 Chr.16: 4359278-4359278 on Build GRCh38
rs759426740 Chr.16: 4359303-4359303 on Build GRCh38
rs778330329 Chr.16: 4359358-4359358 on Build GRCh38
rs1025166224 Chr.16: 4359244-4359244 on Build GRCh38
rs903602013 Chr.16: 4359196-4359196 on Build GRCh38
rs765254955 Chr.16: 4359322-4359322 on Build GRCh38
rs780550916 Chr.16: 4359340-4359340 on Build GRCh38
rs748413691 Chr.16: 4359270-4359270 on Build GRCh38
rs113814539 Chr.16: 4359174-4359175 on Build GRCh38
rs1050548997 Chr.16: 4359173-4359173 on Build GRCh38
rs770430395 Chr.16: 4359357-4359357 on Build GRCh38
rs757701662 Chr.16: 4359248-4359248 on Build GRCh38
rs552059256 Chr.16: 4359332-4359332 on Build GRCh38
rs892651002 Chr.16: 4359230-4359230 on Build GRCh38
rs747462056 Chr.16: 4359342-4359350 on Build GRCh38
rs761770220 Chr.16: 4359277-4359277 on Build GRCh38
rs552144538 Chr.16: 4359164-4359165 on Build GRCh38
rs757304912 Chr.16: 4359315-4359315 on Build GRCh38
rs760876582 Chr.16: 4359312-4359312 on Build GRCh38
rs770358307 Chr.16: 4359301-4359301 on Build GRCh38
rs776798568 Chr.16: 4359313-4359313 on Build GRCh38
rs931624419 Chr.16: 4359154-4359154 on Build GRCh38
rs754013218 Chr.16: 4359314-4359314 on Build GRCh38
rs1052584391 Chr.16: 4359204-4359204 on Build GRCh38
rs758877138 Chr.16: 4359333-4359333 on Build GRCh38
rs755396280 Chr.16: 4359349-4359349 on Build GRCh38
rs779412784 Chr.16: 4359250-4359250 on Build GRCh38
rs199853197 Chr.16: 4359345-4359345 on Build GRCh38
rs868703084 Chr.16: 4359334-4359334 on Build GRCh38
rs775316162 Chr.16: 4359311-4359311 on Build GRCh38
rs767273606 Chr.16: 4359309-4359309 on Build GRCh38
rs982928766 Chr.16: 4359186-4359186 on Build GRCh38
rs192825669 Chr.16: 4359275-4359275 on Build GRCh38
rs750909584 Chr.16: 4359328-4359328 on Build GRCh38
rs377210331 Chr.16: 4359268-4359268 on Build GRCh38
rs764139188 Chr.16: 4359246-4359246 on Build GRCh38

View Full Product Details