Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94988902-94989163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2C9
Entrez Gene ID
1559
Gene Name
cytochrome P450 family 2 subfamily C member 9
Gene Aliases
CPC9, CYP2C, CYP2C10, CYPIIC9, P450IIC9
UniGene
Hs.282624
Gene Chromosome Location
Chr.10: 94905867-94989395 on Build GRCh38
Transcripts
NM_000771.3 XM_017015758.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4451756 c.1397A>G p.N466S Chr.10: 94988952-94988952 on Build GRCh38
921748 c.1399C>A p.L467I Chr.10: 94988954-94988954 on Build GRCh38
1702816 c.1390C>T p.P464S Chr.10: 94988945-94988945 on Build GRCh38
7295249 c.1393A>G p.K465E Chr.10: 94988948-94988948 on Build GRCh38
4990025 c.1368T>C p.F456F Chr.10: 94988923-94988923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867786171 Chr.10: 94988987-94988987 on Build GRCh38
rs890676881 Chr.10: 94989069-94989069 on Build GRCh38
rs760363082 Chr.10: 94989054-94989054 on Build GRCh38
rs760333401 Chr.10: 94988965-94988965 on Build GRCh38
rs9332239 Chr.10: 94989020-94989020 on Build GRCh38
rs759392078 Chr.10: 94988954-94988954 on Build GRCh38
rs9332241 Chr.10: 94989116-94989116 on Build GRCh38
rs376962288 Chr.10: 94988970-94988970 on Build GRCh38
rs943396313 Chr.10: 94989085-94989085 on Build GRCh38
rs112354725 Chr.10: 94988944-94988944 on Build GRCh38
rs756824711 Chr.10: 94988973-94988973 on Build GRCh38
rs922417068 Chr.10: 94989038-94989038 on Build GRCh38
rs961179295 Chr.10: 94989087-94989087 on Build GRCh38
rs757969796 Chr.10: 94989076-94989076 on Build GRCh38
rs9332242 Chr.10: 94989136-94989136 on Build GRCh38
rs771933993 Chr.10: 94989035-94989035 on Build GRCh38
rs767891172 Chr.10: 94989080-94989080 on Build GRCh38
rs1054453493 Chr.10: 94988931-94988931 on Build GRCh38
rs577147873 Chr.10: 94989088-94989088 on Build GRCh38
rs868182778 Chr.10: 94989023-94989023 on Build GRCh38
rs776357745 Chr.10: 94989059-94989059 on Build GRCh38
rs778209778 Chr.10: 94989001-94989007 on Build GRCh38
rs781583846 Chr.10: 94988984-94988984 on Build GRCh38
rs945634152 Chr.10: 94989110-94989110 on Build GRCh38
rs530950257 Chr.10: 94988994-94988994 on Build GRCh38
rs758947599 Chr.10: 94988978-94988978 on Build GRCh38
rs372707224 Chr.10: 94989063-94989063 on Build GRCh38
rs375918804 Chr.10: 94989061-94989061 on Build GRCh38
rs866733675 Chr.10: 94989079-94989079 on Build GRCh38
rs1003261227 Chr.10: 94988985-94988985 on Build GRCh38
rs775537358 Chr.10: 94989052-94989052 on Build GRCh38
rs186465817 Chr.10: 94989094-94989094 on Build GRCh38
rs1053814584 Chr.10: 94989012-94989012 on Build GRCh38
rs748432830 Chr.10: 94988990-94988990 on Build GRCh38
rs752456757 Chr.10: 94988964-94988964 on Build GRCh38
rs376114904 Chr.10: 94988995-94988995 on Build GRCh38
rs745770093 Chr.10: 94989033-94989033 on Build GRCh38
rs141011391 Chr.10: 94988976-94988976 on Build GRCh38
rs767284820 Chr.10: 94988955-94988955 on Build GRCh38
rs749937042 Chr.10: 94989068-94989068 on Build GRCh38
rs752055497 Chr.10: 94988977-94988977 on Build GRCh38
rs151194062 Chr.10: 94988974-94988974 on Build GRCh38
rs906729907 Chr.10: 94989134-94989134 on Build GRCh38
rs749575375 Chr.10: 94989005-94989005 on Build GRCh38
rs763806260 Chr.10: 94988968-94988968 on Build GRCh38
rs774423253 Chr.10: 94989030-94989030 on Build GRCh38
rs1057911 Chr.10: 94988980-94988980 on Build GRCh38
rs879131618 Chr.10: 94989026-94989026 on Build GRCh38
rs9332240 Chr.10: 94989095-94989095 on Build GRCh38

View Full Product Details