Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129098238-129098485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAT
Entrez Gene ID
1384
Gene Name
carnitine O-acetyltransferase
Gene Aliases
CAT1
UniGene
Hs.12068
Gene Chromosome Location
Chr.9: 129094794-129110791 on Build GRCh38
Transcripts
NM_000755.3 NM_001257363.1 XM_005251708.3 XM_017014274.1 XM_017014275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
752581 c.1242C>T p.F414F Chr.9: 129098335-129098335 on Build GRCh38
1489719 c.1245C>T p.H415H Chr.9: 129098332-129098332 on Build GRCh38
6253961 c.1284C>A p.S428R Chr.9: 129098293-129098293 on Build GRCh38
1636056 c.1213C>A p.Q405K Chr.9: 129098364-129098364 on Build GRCh38
376968 c.1253G>A p.G418E Chr.9: 129098324-129098324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112769430 Chr.9: 129098429-129098429 on Build GRCh38
rs1041149091 Chr.9: 129098423-129098423 on Build GRCh38
rs17486094 Chr.9: 129098446-129098446 on Build GRCh38
rs777563979 Chr.9: 129098330-129098330 on Build GRCh38
rs755854715 Chr.9: 129098275-129098275 on Build GRCh38
rs777509069 Chr.9: 129098421-129098421 on Build GRCh38
rs764664238 Chr.9: 129098409-129098409 on Build GRCh38
rs946600875 Chr.9: 129098305-129098305 on Build GRCh38
rs758296270 Chr.9: 129098415-129098415 on Build GRCh38
rs535503674 Chr.9: 129098408-129098408 on Build GRCh38
rs774887997 Chr.9: 129098379-129098379 on Build GRCh38
rs888409744 Chr.9: 129098445-129098445 on Build GRCh38
rs1048410175 Chr.9: 129098444-129098444 on Build GRCh38
rs182284225 Chr.9: 129098346-129098346 on Build GRCh38
rs753447240 Chr.9: 129098287-129098287 on Build GRCh38
rs375358081 Chr.9: 129098398-129098398 on Build GRCh38
rs750039829 Chr.9: 129098266-129098266 on Build GRCh38
rs201109466 Chr.9: 129098317-129098317 on Build GRCh38
rs771187276 Chr.9: 129098345-129098345 on Build GRCh38
rs889820107 Chr.9: 129098384-129098384 on Build GRCh38
rs150028237 Chr.9: 129098306-129098306 on Build GRCh38
rs569811614 Chr.9: 129098365-129098365 on Build GRCh38
rs748037993 Chr.9: 129098314-129098314 on Build GRCh38
rs773176599 Chr.9: 129098335-129098335 on Build GRCh38
rs934112186 Chr.9: 129098374-129098374 on Build GRCh38
rs750895770 Chr.9: 129098337-129098341 on Build GRCh38
rs752628842 Chr.9: 129098410-129098410 on Build GRCh38
rs753282397 Chr.9: 129098403-129098403 on Build GRCh38
rs374143667 Chr.9: 129098347-129098347 on Build GRCh38
rs566951989 Chr.9: 129098295-129098295 on Build GRCh38
rs368706124 Chr.9: 129098378-129098378 on Build GRCh38
rs558520570 Chr.9: 129098393-129098393 on Build GRCh38
rs774333847 Chr.9: 129098267-129098267 on Build GRCh38
rs923754832 Chr.9: 129098373-129098374 on Build GRCh38
rs372331656 Chr.9: 129098383-129098383 on Build GRCh38
rs770200532 Chr.9: 129098349-129098349 on Build GRCh38
rs140551503 Chr.9: 129098283-129098283 on Build GRCh38
rs200167828 Chr.9: 129098375-129098375 on Build GRCh38
rs747189121 Chr.9: 129098343-129098343 on Build GRCh38
rs929992501 Chr.9: 129098440-129098440 on Build GRCh38

View Full Product Details