Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154861663-154861924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F8
Entrez Gene ID
2157
Gene Name
coagulation factor VIII
Gene Aliases
AHF, DXS1253E, F8B, F8C, FVIII, HEMA
UniGene
Hs.632836
Gene Chromosome Location
Chr.X: 154835788-155022723 on Build GRCh38
Transcripts
NM_000132.3 NM_019863.2

SNP Detail

SNP ID Chromosome Location
rs782548763 Chr.X: 154861783-154861783 on Build GRCh38
rs387906464 Chr.X: 154861742-154861742 on Build GRCh38
rs370137203 Chr.X: 154861711-154861711 on Build GRCh38
rs782428789 Chr.X: 154861740-154861740 on Build GRCh38
rs782627131 Chr.X: 154861789-154861789 on Build GRCh38
rs782433673 Chr.X: 154861690-154861690 on Build GRCh38
rs782198570 Chr.X: 154861818-154861818 on Build GRCh38
rs137852355 Chr.X: 154861759-154861759 on Build GRCh38
rs782098979 Chr.X: 154861866-154861866 on Build GRCh38
rs782407735 Chr.X: 154861850-154861850 on Build GRCh38
rs782567782 Chr.X: 154861808-154861808 on Build GRCh38
rs781957923 Chr.X: 154861862-154861862 on Build GRCh38
rs1800296 Chr.X: 154861799-154861799 on Build GRCh38
rs781838306 Chr.X: 154861705-154861705 on Build GRCh38
rs782727913 Chr.X: 154861697-154861697 on Build GRCh38
rs920493459 Chr.X: 154861713-154861713 on Build GRCh38
rs375400068 Chr.X: 154861778-154861778 on Build GRCh38
rs146151112 Chr.X: 154861817-154861817 on Build GRCh38
rs137852468 Chr.X: 154861810-154861810 on Build GRCh38
rs782036886 Chr.X: 154861858-154861858 on Build GRCh38
rs200114987 Chr.X: 154861773-154861773 on Build GRCh38
rs782394042 Chr.X: 154861861-154861861 on Build GRCh38
rs137852358 Chr.X: 154861758-154861758 on Build GRCh38
rs1008606692 Chr.X: 154861860-154861860 on Build GRCh38
rs377399008 Chr.X: 154861693-154861693 on Build GRCh38
rs782366441 Chr.X: 154861805-154861805 on Build GRCh38
rs781804352 Chr.X: 154861897-154861897 on Build GRCh38
rs782244990 Chr.X: 154861876-154861876 on Build GRCh38
rs201870876 Chr.X: 154861745-154861745 on Build GRCh38
rs139144797 Chr.X: 154861859-154861859 on Build GRCh38

View Full Product Details