Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61419965-61420204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF7
Entrez Gene ID
79869
Gene Name
cleavage and polyadenylation specific factor 7
Gene Aliases
CFIm59
UniGene
Hs.718984
Gene Chromosome Location
Chr.11: 61402648-61429992 on Build GRCh38
Transcripts
NM_001136040.2 NM_001142565.1 NM_024811.3 XM_005274298.4 XM_005274299.4 XM_005274303.4 XM_011545257.2 XM_011545258.2 XM_011545259.2 XM_011545260.2 XM_011545261.2 XM_011545262.2 XM_011545263.2 XM_017018345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1638899 c.384T>C p.A128A Chr.11: 61420088-61420088 on Build GRCh38
1638898 c.440G>T p.G147V Chr.11: 61420032-61420032 on Build GRCh38
4812856 c.634G>A p.E212K Chr.11: 61419967-61419967 on Build GRCh38
4674482 c.439G>T p.G147W Chr.11: 61420033-61420033 on Build GRCh38
6634922 c.613C>T p.R205W Chr.11: 61419988-61419988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776120059 Chr.11: 61420146-61420146 on Build GRCh38
rs781139594 Chr.11: 61420070-61420070 on Build GRCh38
rs780092071 Chr.11: 61420049-61420049 on Build GRCh38
rs764427271 Chr.11: 61420000-61420000 on Build GRCh38
rs748868804 Chr.11: 61420130-61420130 on Build GRCh38
rs747894968 Chr.11: 61420076-61420076 on Build GRCh38
rs754888784 Chr.11: 61420058-61420058 on Build GRCh38
rs138161429 Chr.11: 61420004-61420004 on Build GRCh38
rs758380572 Chr.11: 61420044-61420044 on Build GRCh38
rs751439980 Chr.11: 61420055-61420055 on Build GRCh38
rs200063140 Chr.11: 61420118-61420118 on Build GRCh38
rs369351140 Chr.11: 61420042-61420042 on Build GRCh38
rs554088161 Chr.11: 61420012-61420012 on Build GRCh38
rs755852770 Chr.11: 61420098-61420098 on Build GRCh38
rs373828067 Chr.11: 61419986-61419986 on Build GRCh38
rs920565609 Chr.11: 61420011-61420011 on Build GRCh38
rs911673949 Chr.11: 61420162-61420162 on Build GRCh38
rs542849832 Chr.11: 61420141-61420141 on Build GRCh38
rs914981244 Chr.11: 61420007-61420007 on Build GRCh38
rs199690496 Chr.11: 61420021-61420021 on Build GRCh38
rs765399178 Chr.11: 61420030-61420030 on Build GRCh38
rs1045495184 Chr.11: 61420071-61420071 on Build GRCh38
rs906435618 Chr.11: 61420134-61420134 on Build GRCh38
rs985975504 Chr.11: 61420142-61420142 on Build GRCh38

View Full Product Details