Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75762667-75762904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALK1
Entrez Gene ID
2584
Gene Name
galactokinase 1
Gene Aliases
GALK, GK1, HEL-S-19
UniGene
Hs.407966
Gene Chromosome Location
Chr.17: 75757937-75765199 on Build GRCh38
Transcripts
NM_000154.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3522310 c.765C>T p.L255L Chr.17: 75762732-75762732 on Build GRCh38
6668937 c.767G>A p.R256Q Chr.17: 75762730-75762730 on Build GRCh38
7369149 c.746C>T p.A249V Chr.17: 75762751-75762751 on Build GRCh38
4686238 c.671A>G p.N224S Chr.17: 75762826-75762826 on Build GRCh38
4686237 c.721C>T p.R241C Chr.17: 75762776-75762776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146095595 Chr.17: 75762732-75762732 on Build GRCh38
rs762774725 Chr.17: 75762776-75762776 on Build GRCh38
rs140053358 Chr.17: 75762750-75762750 on Build GRCh38
rs770716297 Chr.17: 75762862-75762862 on Build GRCh38
rs368895044 Chr.17: 75762808-75762808 on Build GRCh38
rs760894170 Chr.17: 75762880-75762880 on Build GRCh38
rs575139300 Chr.17: 75762781-75762781 on Build GRCh38
rs752685775 Chr.17: 75762706-75762706 on Build GRCh38
rs374114654 Chr.17: 75762814-75762814 on Build GRCh38
rs759699355 Chr.17: 75762702-75762702 on Build GRCh38
rs201939520 Chr.17: 75762852-75762852 on Build GRCh38
rs780306197 Chr.17: 75762836-75762836 on Build GRCh38
rs758885920 Chr.17: 75762827-75762827 on Build GRCh38
rs148557440 Chr.17: 75762855-75762855 on Build GRCh38
rs904702679 Chr.17: 75762749-75762749 on Build GRCh38
rs748015014 Chr.17: 75762760-75762760 on Build GRCh38
rs149825190 Chr.17: 75762751-75762751 on Build GRCh38
rs773559496 Chr.17: 75762787-75762787 on Build GRCh38
rs754329715 Chr.17: 75762728-75762728 on Build GRCh38
rs746635166 Chr.17: 75762839-75762839 on Build GRCh38
rs373103005 Chr.17: 75762775-75762775 on Build GRCh38
rs773178321 Chr.17: 75762857-75762857 on Build GRCh38
rs368904895 Chr.17: 75762754-75762754 on Build GRCh38
rs756263074 Chr.17: 75762711-75762711 on Build GRCh38
rs754593748 Chr.17: 75762840-75762840 on Build GRCh38
rs376790302 Chr.17: 75762731-75762731 on Build GRCh38
rs768818717 Chr.17: 75762883-75762883 on Build GRCh38
rs369906636 Chr.17: 75762856-75762856 on Build GRCh38
rs774107830 Chr.17: 75762753-75762753 on Build GRCh38
rs561544412 Chr.17: 75762762-75762762 on Build GRCh38
rs981970282 Chr.17: 75762825-75762825 on Build GRCh38
rs764167059 Chr.17: 75762794-75762794 on Build GRCh38
rs767790909 Chr.17: 75762703-75762703 on Build GRCh38
rs372321234 Chr.17: 75762730-75762730 on Build GRCh38
rs544282036 Chr.17: 75762815-75762815 on Build GRCh38
rs376343759 Chr.17: 75762779-75762779 on Build GRCh38
rs897784159 Chr.17: 75762772-75762772 on Build GRCh38
rs1030442171 Chr.17: 75762704-75762704 on Build GRCh38
rs369795225 Chr.17: 75762685-75762685 on Build GRCh38
rs369392713 Chr.17: 75762795-75762795 on Build GRCh38
rs886053420 Chr.17: 75762710-75762710 on Build GRCh38
rs145837971 Chr.17: 75762778-75762778 on Build GRCh38
rs764099539 Chr.17: 75762726-75762726 on Build GRCh38
rs866856435 Chr.17: 75762757-75762757 on Build GRCh38
rs749242779 Chr.17: 75762858-75762858 on Build GRCh38
rs541580127 Chr.17: 75762755-75762755 on Build GRCh38
rs980603321 Chr.17: 75762789-75762789 on Build GRCh38
rs371785693 Chr.17: 75762872-75762872 on Build GRCh38
rs746480367 Chr.17: 75762752-75762752 on Build GRCh38

View Full Product Details