Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169515403-169515676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
UniGene
Hs.30054
Gene Chromosome Location
Chr.1: 169511954-169586632 on Build GRCh38
Transcripts
NM_000130.4 XM_017000660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7086130 c.6453G>C p.K2151N Chr.1: 169515519-169515519 on Build GRCh38
7089630 c.6480G>T p.Q2160H Chr.1: 169515492-169515492 on Build GRCh38
899252 c.6401C>T p.T2134M Chr.1: 169515571-169515571 on Build GRCh38
4808077 c.6346-10A>G p.? Chr.1: 169515636-169515636 on Build GRCh38
3863464 c.6418G>A p.G2140S Chr.1: 169515554-169515554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369720229 Chr.1: 169515448-169515448 on Build GRCh38
rs200777907 Chr.1: 169515512-169515512 on Build GRCh38
rs138021584 Chr.1: 169515483-169515483 on Build GRCh38
rs767524616 Chr.1: 169515569-169515569 on Build GRCh38
rs199601865 Chr.1: 169515571-169515571 on Build GRCh38
rs889356518 Chr.1: 169515501-169515501 on Build GRCh38
rs144486119 Chr.1: 169515570-169515570 on Build GRCh38
rs746504481 Chr.1: 169515645-169515645 on Build GRCh38
rs1032493902 Chr.1: 169515435-169515435 on Build GRCh38
rs774445274 Chr.1: 169515547-169515547 on Build GRCh38
rs201920792 Chr.1: 169515520-169515520 on Build GRCh38
rs773618429 Chr.1: 169515513-169515513 on Build GRCh38
rs756075594 Chr.1: 169515441-169515441 on Build GRCh38
rs770101014 Chr.1: 169515440-169515443 on Build GRCh38
rs112942876 Chr.1: 169515432-169515432 on Build GRCh38
rs779457427 Chr.1: 169515637-169515637 on Build GRCh38
rs753754441 Chr.1: 169515602-169515602 on Build GRCh38
rs757104503 Chr.1: 169515612-169515612 on Build GRCh38
rs369788298 Chr.1: 169515635-169515635 on Build GRCh38
rs780625444 Chr.1: 169515456-169515456 on Build GRCh38
rs764090739 Chr.1: 169515598-169515598 on Build GRCh38
rs771496715 Chr.1: 169515495-169515495 on Build GRCh38
rs892889950 Chr.1: 169515565-169515565 on Build GRCh38
rs9332701 Chr.1: 169515529-169515529 on Build GRCh38
rs6004 Chr.1: 169515630-169515630 on Build GRCh38
rs1002803558 Chr.1: 169515434-169515434 on Build GRCh38
rs576975311 Chr.1: 169515488-169515488 on Build GRCh38
rs778713126 Chr.1: 169515629-169515629 on Build GRCh38
rs775001649 Chr.1: 169515511-169515511 on Build GRCh38
rs377723406 Chr.1: 169515588-169515588 on Build GRCh38
rs759734191 Chr.1: 169515552-169515552 on Build GRCh38
rs757010994 Chr.1: 169515468-169515468 on Build GRCh38
rs976415610 Chr.1: 169515596-169515596 on Build GRCh38

View Full Product Details