Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42154623-42154874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3M2
Entrez Gene ID
10947
Gene Name
adaptor related protein complex 3 mu 2 subunit
Gene Aliases
AP47B, CLA20, P47B
UniGene
Hs.654529
Gene Chromosome Location
Chr.8: 42152946-42171183 on Build GRCh38
Transcripts
NM_001134296.1 NM_006803.3 XM_017012977.1 XM_017012978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5037811 c.151A>C p.T51P Chr.8: 42154838-42154838 on Build GRCh38
6379090 c.179A>T p.Y60F Chr.8: 42154866-42154866 on Build GRCh38
6276849 c.11G>T p.S4I Chr.8: 42154698-42154698 on Build GRCh38
1635801 c.4A>G p.I2V Chr.8: 42154691-42154691 on Build GRCh38
3649311 c.25A>T p.N9Y Chr.8: 42154712-42154712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773206834 Chr.8: 42154648-42154648 on Build GRCh38
rs771006544 Chr.8: 42154780-42154780 on Build GRCh38
rs528347317 Chr.8: 42154652-42154652 on Build GRCh38
rs753042418 Chr.8: 42154838-42154838 on Build GRCh38
rs760422293 Chr.8: 42154660-42154660 on Build GRCh38
rs1026602662 Chr.8: 42154822-42154822 on Build GRCh38
rs567884622 Chr.8: 42154681-42154681 on Build GRCh38
rs374433477 Chr.8: 42154649-42154649 on Build GRCh38
rs371712957 Chr.8: 42154740-42154740 on Build GRCh38
rs200545949 Chr.8: 42154695-42154695 on Build GRCh38
rs369061760 Chr.8: 42154809-42154809 on Build GRCh38
rs552895671 Chr.8: 42154800-42154800 on Build GRCh38
rs764961587 Chr.8: 42154802-42154802 on Build GRCh38
rs769995342 Chr.8: 42154763-42154763 on Build GRCh38
rs911059208 Chr.8: 42154721-42154721 on Build GRCh38
rs758994283 Chr.8: 42154686-42154686 on Build GRCh38
rs201026159 Chr.8: 42154829-42154829 on Build GRCh38
rs749301256 Chr.8: 42154779-42154779 on Build GRCh38
rs752019367 Chr.8: 42154833-42154833 on Build GRCh38
rs775315240 Chr.8: 42154784-42154784 on Build GRCh38
rs756374378 Chr.8: 42154849-42154849 on Build GRCh38
rs762400472 Chr.8: 42154827-42154827 on Build GRCh38
rs369414381 Chr.8: 42154716-42154716 on Build GRCh38
rs775400442 Chr.8: 42154659-42154659 on Build GRCh38
rs770178462 Chr.8: 42154679-42154679 on Build GRCh38
rs777884964 Chr.8: 42154764-42154764 on Build GRCh38
rs764908425 Chr.8: 42154682-42154682 on Build GRCh38
rs1048198978 Chr.8: 42154828-42154828 on Build GRCh38
rs772861829 Chr.8: 42154673-42154673 on Build GRCh38
rs1039117920 Chr.8: 42154783-42154783 on Build GRCh38
rs149242459 Chr.8: 42154791-42154791 on Build GRCh38
rs199664300 Chr.8: 42154792-42154792 on Build GRCh38
rs923915092 Chr.8: 42154671-42154671 on Build GRCh38
rs200178239 Chr.8: 42154711-42154711 on Build GRCh38
rs372555268 Chr.8: 42154834-42154834 on Build GRCh38
rs773828482 Chr.8: 42154700-42154700 on Build GRCh38
rs140456285 Chr.8: 42154717-42154717 on Build GRCh38
rs35071361 Chr.8: 42154668-42154668 on Build GRCh38
rs942523181 Chr.8: 42154762-42154762 on Build GRCh38
rs749941452 Chr.8: 42154685-42154685 on Build GRCh38

View Full Product Details