Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 105617952-105618181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKTN
Entrez Gene ID
2218
Gene Name
fukutin
Gene Aliases
CMD1X, FCMD, LGMD2M, MDDGA4, MDDGB4, MDDGC4
UniGene
Hs.55777
Gene Chromosome Location
Chr.9: 105558117-105655950 on Build GRCh38
Transcripts
NM_001079802.1 NM_001198963.1 NM_006731.2 XM_006717014.2 XM_011518368.2 XM_011518369.2 XM_011518370.2 XM_011518371.2 XM_011518373.2 XM_011518374.2 XM_011518375.2 XM_011518376.2 XM_011518378.2 XM_011518379.2 XM_011518381.2 XM_011518387.2 XM_011518389.2 XM_011518390.2 XM_011518391.2 XM_017014462.1 XM_017014463.1 XM_017014464.1 XM_017014465.1 XM_017014466.1 XM_017014467.1 XM_017014468.1 XM_017014469.1 XM_017014470.1 XM_017014471.1 XM_017014472.1 XM_017014473.1 XM_017014474.1 XM_017014475.1 XM_017014476.1 XM_017014477.1 XM_017014478.1 XM_017014479.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467555 c.919C>T p.R307* Chr.9: 105617967-105617967 on Build GRCh38
1103574 c.949G>T p.D317Y Chr.9: 105617997-105617997 on Build GRCh38
3652621 c.1006C>T p.Q336* Chr.9: 105618054-105618054 on Build GRCh38
6366241 c.976G>T p.D326Y Chr.9: 105618024-105618024 on Build GRCh38
4163187 c.1026C>A p.L342L Chr.9: 105618074-105618074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774410536 Chr.9: 105618023-105618023 on Build GRCh38
rs75946934 Chr.9: 105618150-105618150 on Build GRCh38
rs777297548 Chr.9: 105618121-105618124 on Build GRCh38
rs1018122957 Chr.9: 105618026-105618026 on Build GRCh38
rs750787127 Chr.9: 105618085-105618085 on Build GRCh38
rs992936408 Chr.9: 105618148-105618148 on Build GRCh38
rs748169818 Chr.9: 105618135-105618135 on Build GRCh38
rs764441249 Chr.9: 105618070-105618070 on Build GRCh38
rs146967918 Chr.9: 105618071-105618071 on Build GRCh38
rs200801909 Chr.9: 105618109-105618109 on Build GRCh38
rs780980837 Chr.9: 105618130-105618130 on Build GRCh38
rs915268305 Chr.9: 105618134-105618134 on Build GRCh38
rs752721354 Chr.9: 105618057-105618057 on Build GRCh38
rs17309806 Chr.9: 105618074-105618074 on Build GRCh38
rs746456728 Chr.9: 105618116-105618116 on Build GRCh38
rs769691253 Chr.9: 105618020-105618020 on Build GRCh38
rs951073727 Chr.9: 105618145-105618145 on Build GRCh38
rs141120187 Chr.9: 105618000-105618000 on Build GRCh38
rs886044385 Chr.9: 105618069-105618069 on Build GRCh38
rs759791267 Chr.9: 105618025-105618025 on Build GRCh38
rs773018262 Chr.9: 105618138-105618138 on Build GRCh38
rs970907026 Chr.9: 105617990-105617990 on Build GRCh38
rs924159503 Chr.9: 105618053-105618053 on Build GRCh38
rs768352058 Chr.9: 105618129-105618129 on Build GRCh38
rs118064041 Chr.9: 105618136-105618136 on Build GRCh38
rs767753943 Chr.9: 105618028-105618028 on Build GRCh38
rs760756067 Chr.9: 105618058-105618058 on Build GRCh38
rs532960237 Chr.9: 105618151-105618151 on Build GRCh38
rs751122944 Chr.9: 105618063-105618063 on Build GRCh38

View Full Product Details