Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33700601-33700830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf91
Entrez Gene ID
100131378
Gene Name
chromosome 11 open reading frame 91
Gene Aliases
-
UniGene
Hs.709466
Gene Chromosome Location
Chr.11: 33698108-33703026 on Build GRCh38
Transcripts
NM_001166692.1 XM_017017053.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5952549 c.117C>A p.F39L Chr.11: 33700624-33700624 on Build GRCh38
6189553 c.138C>A p.F46L Chr.11: 33700603-33700603 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs941243332 Chr.11: 33700665-33700665 on Build GRCh38
rs560695528 Chr.11: 33700725-33700725 on Build GRCh38
rs1045437498 Chr.11: 33700713-33700713 on Build GRCh38
rs767954708 Chr.11: 33700763-33700763 on Build GRCh38
rs964244556 Chr.11: 33700637-33700637 on Build GRCh38
rs902906095 Chr.11: 33700719-33700719 on Build GRCh38
rs763217367 Chr.11: 33700638-33700638 on Build GRCh38
rs529730497 Chr.11: 33700778-33700778 on Build GRCh38
rs930289271 Chr.11: 33700643-33700643 on Build GRCh38
rs949922621 Chr.11: 33700712-33700712 on Build GRCh38
rs1028856694 Chr.11: 33700672-33700672 on Build GRCh38
rs886173596 Chr.11: 33700647-33700647 on Build GRCh38
rs920182684 Chr.11: 33700642-33700642 on Build GRCh38
rs995971261 Chr.11: 33700670-33700670 on Build GRCh38
rs753252216 Chr.11: 33700774-33700774 on Build GRCh38
rs967797192 Chr.11: 33700798-33700798 on Build GRCh38
rs1048798899 Chr.11: 33700646-33700646 on Build GRCh38
rs976824809 Chr.11: 33700639-33700639 on Build GRCh38
rs541996192 Chr.11: 33700663-33700663 on Build GRCh38
rs1040545767 Chr.11: 33700656-33700656 on Build GRCh38
rs755393600 Chr.11: 33700707-33700707 on Build GRCh38
rs773246914 Chr.11: 33700625-33700625 on Build GRCh38
rs1011512925 Chr.11: 33700748-33700748 on Build GRCh38
rs577746897 Chr.11: 33700721-33700721 on Build GRCh38
rs751917753 Chr.11: 33700702-33700702 on Build GRCh38
rs940422019 Chr.11: 33700649-33700649 on Build GRCh38

View Full Product Details