Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 165289797-165290028 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf118
Entrez Gene ID
168090
Gene Name
chromosome 6 open reading frame 118
Gene Aliases
bA85G2.1, dJ416F21.2
UniGene
Hs.144734
Gene Chromosome Location
Chr.6: 165279664-165309647 on Build GRCh38
Transcripts
NM_144980.3 XM_005266838.2 XM_011535509.2 XM_011535510.2 XM_011535511.2 XM_011535512.2 XM_011535513.2 XM_017010323.1 XM_017010324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5946937 c.1270G>A p.D424N Chr.6: 165289918-165289918 on Build GRCh38
4618576 c.1214A>C p.E405A Chr.6: 165289974-165289974 on Build GRCh38
1315766 c.1226T>C p.L409P Chr.6: 165289962-165289962 on Build GRCh38
6616394 c.1275C>A p.I425I Chr.6: 165289913-165289913 on Build GRCh38
3622721 c.1175T>C p.V392A Chr.6: 165290013-165290013 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs73022263 Chr.6: 165289964-165289964 on Build GRCh38
rs757750550 Chr.6: 165289948-165289948 on Build GRCh38
rs754636643 Chr.6: 165289843-165289843 on Build GRCh38
rs747936548 Chr.6: 165289954-165289955 on Build GRCh38
rs568745121 Chr.6: 165289864-165289864 on Build GRCh38
rs528908031 Chr.6: 165289935-165289935 on Build GRCh38
rs759221879 Chr.6: 165289910-165289910 on Build GRCh38
rs773039565 Chr.6: 165289977-165289977 on Build GRCh38
rs368161744 Chr.6: 165289874-165289874 on Build GRCh38
rs922066459 Chr.6: 165289914-165289914 on Build GRCh38
rs781167038 Chr.6: 165289881-165289881 on Build GRCh38
rs752077898 Chr.6: 165289944-165289944 on Build GRCh38
rs770840353 Chr.6: 165289980-165289980 on Build GRCh38
rs773166200 Chr.6: 165289893-165289893 on Build GRCh38
rs776140484 Chr.6: 165289990-165289990 on Build GRCh38
rs780280648 Chr.6: 165289958-165289958 on Build GRCh38
rs745943743 Chr.6: 165289952-165289952 on Build GRCh38
rs779321262 Chr.6: 165289844-165289844 on Build GRCh38
rs202221904 Chr.6: 165289899-165289899 on Build GRCh38
rs780184501 Chr.6: 165289953-165289954 on Build GRCh38
rs770090641 Chr.6: 165289901-165289901 on Build GRCh38
rs760645691 Chr.6: 165289898-165289898 on Build GRCh38
rs753051233 Chr.6: 165289921-165289921 on Build GRCh38
rs753345868 Chr.6: 165289902-165289902 on Build GRCh38
rs771949813 Chr.6: 165289890-165289890 on Build GRCh38
rs758906972 Chr.6: 165289856-165289856 on Build GRCh38
rs550591403 Chr.6: 165289835-165289835 on Build GRCh38
rs758853353 Chr.6: 165289926-165289926 on Build GRCh38
rs747900020 Chr.6: 165289962-165289962 on Build GRCh38
rs774131750 Chr.6: 165289883-165289883 on Build GRCh38
rs781332152 Chr.6: 165289949-165289949 on Build GRCh38
rs575992476 Chr.6: 165289937-165289937 on Build GRCh38
rs756304210 Chr.6: 165289953-165289953 on Build GRCh38
rs775309888 Chr.6: 165289998-165289998 on Build GRCh38
rs746937707 Chr.6: 165289978-165289978 on Build GRCh38
rs778382231 Chr.6: 165289932-165289932 on Build GRCh38
rs373965245 Chr.6: 165289853-165289853 on Build GRCh38
rs762731673 Chr.6: 165290000-165290000 on Build GRCh38
rs989277623 Chr.6: 165289927-165289927 on Build GRCh38
rs753179641 Chr.6: 165289848-165289848 on Build GRCh38
rs764893135 Chr.6: 165289995-165289995 on Build GRCh38
rs182643871 Chr.6: 165289882-165289882 on Build GRCh38
rs761612484 Chr.6: 165289885-165289885 on Build GRCh38
rs764800248 Chr.6: 165289920-165289920 on Build GRCh38
rs758980537 Chr.6: 165289991-165289991 on Build GRCh38
rs771071816 Chr.6: 165289876-165289876 on Build GRCh38
rs899213061 Chr.6: 165289834-165289834 on Build GRCh38

View Full Product Details